Skip to Content
Merck
All Photos(1)

Key Documents

MSPS004

Sigma-Aldrich

SILuPep C-peptide

synthetic, SIL MS Peptide Standard, ¹³C labeled

Synonym(s):

Serotransferrin

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€245.00

€245.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 μG
€245.00

About This Item

UNSPSC Code:
23201100
NACRES:
NA.24

€245.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

≥95% (HPLC)

form

lyophilized

color

off-white

shipped in

ambient

storage temp.

−20°C

Related Categories

General description

SILuPep C-peptide is a synthetic, stable isotope-labeled human peptide.

Analysis Note

Sequence
EAEDLQVGQVELGGGPGAGSLQPLA-Leu(U13C6)-EGS-Leu(U13C6)-Q
Terminal modifications: N-Term: [H] C-Term: [OH]
Length (residues): 31

Label Incorporation
>= 98% as determined by mass spectrometry

Legal Information

SILu is a trademark of Sigma-Aldrich Co. LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service