Skip to Content

MSPS002

SILuPep GLP-1 active

synthetic, SIL MS Peptide Standard, ¹³C labeled

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NA.24
UNSPSC Code:
23201100
Assay:
≥95% (HPLC)
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥95% (HPLC)

form

lyophilized

color

off-white

shipped in

ambient

storage temp.

−20°C

Analysis Note

General Description
SILuPep GLP-1 Active is a synthetic, stable isotope-labeled human peptide.

Sequence
HAEGTFTSDVSSY-Leu(U13C6)-EGQAAKEFIAW-Leu(U13C6)-VKGR-NH2
Terminal modifications: N-Term: [H]
Length (residues): 30

Label Incorporation
>= 98% as determined by mass spectrometry

Legal Information

SILu is a trademark of Sigma-Aldrich Co. LLC


pictograms

Health hazardCorrosion

signalword

Danger

Hazard Classifications

Eye Dam. 1 - Repr. 2 - Skin Corr. 1A

Storage Class

8A - Combustible corrosive hazardous materials

flash_point_f

Not applicable

flash_point_c

Not applicable

supp_hazards


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

75-05-8

CAS No.


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library