Skip to Content
Merck
All Photos(1)

Key Documents

MSKT0003

Sigma-Aldrich

IA-MS Immunoaffinity Mass Spectrometry Kit - Streptavidin

96 Well Strip Plate with SOLu-Trypsin

Synonym(s):

Target protein enrichment and quantification from complex background by mass spectrometry

Sign Into View Organizational & Contract Pricing

Select a Size

1 KIT
€559.00

€559.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 KIT
€559.00

About This Item

UNSPSC Code:
41115859
NACRES:
NA.41

€559.00


Check Cart for Availability

Request a Bulk Order

form

solid

suitability

suitable for mass spectrometry

shipped in

wet ice

storage temp.

2-8°C

General description

The Streptavidin IA-MS Immunoaffinity Mass Spectrometry Kit is a 96-well platform enabling robust, high-throughput quantification of proteins in complex backgrounds by LC-MS/MS. Users provide their own biotinylated capture molecule and internal standard (available separately, internal standards may be SIL peptides or preferably full length SIL protein) and the kit contains all other components needed for in-well enrichment and rapid trypsin digestion of samples. The plate format consists of 8-well strips allowing for partial runs, and avoids the high cost and potential LC column blockage associated with bead-based enrichment formats. The entire workflow can be performed in under 5 hours, resulting in digests ready for injection with no SPE or other cleanup required.

Application

Target protein enrichment and quantification from complex background by mass spectrometry.

Features and Benefits

  • Entire workflow including digest completed in under 5 hours, such that samples are injection ready at the end of the work day
  • 96-well plate format has potential for automation in a liquid-handling system
  • Bead-free system circumvents messy clean-up and eliminates risk of column clogging
  • Plate strips allow user to run as many or as few samples as desired

Packaging

Box with foam insert containing kit components.

Components

Streptavidin High Binding Capacity Coated Plate S2577-1EA
SOLu-Trypsin EMS0004-100UG
MS Denaturation Solution EMS00010-10ML
Rapid Trypsin Digestion Buffer EMS0009-30ML
Tris Buffered Saline with TWEEN® 20 (TBST)powder, pH 8.0 T9039
Tris Buffered Saline (TBS) powder, pH 8.0 T6664
EZ-Pierce plate seal Z721581-1EA

See datasheet for more information.

Legal Information

EZ-Pierce is a trademark of Excel Scientific, Inc.
TWEEN is a registered trademark of Croda International PLC

Kit Components Only

Product No.
Description

  • Rapid Trypsin Digestion Buffer 30 mL

  • MS Denaturation Solution

Kit Components Also Available Separately

Product No.
Description
SDS

  • S2577Streptavidin High Capacity Coated Plates, 8 well strip, clear 1 plateSDS

  • EMS0004SOLu-Trypsin, recombinant, expressed in Pichia pastoris, Proteomics Grade, liquid 4 ´ 100 mL (1 mg/mL)SDS

  • S2577Streptavidin High Capacity Coated Plates, 8 well strip, clear 1 packetSDS

  • T6664Tris Buffered Saline, pH 8.0, powderSDS

  • Z721581EZ-Pierce films, Excel Scientific EZP-100, non-sterile 4 filmsSDS

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 3 - STOT SE 1

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

75.2 °F

Flash Point(C)

24 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service