Skip to Content
Merck
All Photos(1)

Key Documents

MAK305

Sigma-Aldrich

CETP RP Activity Assay

(Supplied by Roar Biomedical, Inc.)

Sign Into View Organizational & Contract Pricing

Select a Size

1 KIT
€680.00

€680.00


Usually ships in 2 weeks.

Request a Bulk Order

Select a Size

Change View
1 KIT
€680.00

About This Item

UNSPSC Code:
12161503
NACRES:
NA.84

€680.00


Usually ships in 2 weeks.

Request a Bulk Order

usage

sufficient for 100 fluorometric tests

application(s)

pharmaceutical

detection method

fluorometric

relevant disease(s)

cardiovascular diseases

storage temp.

2-8°C

General description

Cholesteryl ester transfer protein (CETP) is present in normal human plasma and transfers neutral lipids from high density lipoproteins (HDL) to very low density lipoprotein (VLDL) and low density lipoprotein (LDL). CETP plays an important role in lipoprotein metabolism and influences the reverse cholesterol transport pathway.

Features and Benefits

Synthetic assay components support manipulation for MOA studies and SAR work. Unusually high signal-to-background with linearity. Assay components stable for up to 1 year. High DMSO tolerance.

Suitability

Suitable for high-throughput screening (HTS), mechanism of action (MOA) studies, and structure-activity relationship (SAR) work in CETP sources.

Principle

The CETP RP Activity Assay uses a proprietary substrate that enables the detection of CETP-mediated neutral lipid mass transfer. The method is useful for measuring CETP activity in recombinant protein (RP) or purified CETP samples and has a high DMSO tolerance. The transfer activity results in an increase in fluorescence intensity (λEx = 465 nm/λEm = 535 nm). With recombinant CETP as the CETP source, the assay is linear over a wide range of CETP concentrations and provides very high signal-to-background values. Typical Z factor: ∼0.9

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service