The product MAK106 uses linear regression fitting. The standard curve is prepared based on the donor particle concentration indicated on the label, which is batch-specific. The standard curve in table 2 is representative and for example purposes only. It's important to note that the concentrations may vary. The pmole values are calculated through dimension analysis using the amount of donor particle solution used and any subsequent dilution. For example, the most concentrated standard can be calculated by considering the volume of the donor particle. In this case, the most concentrated standard has 130 pmoles. Each subsequent dilution is two times less concentrated, resulting in concentrations of 65, 32.5, 16.25, and 8.1 pmoles.
MAK106
CETP Activity Assay Kit
Supplied by Roar Biomedical, Inc.
Synonym(s):
Cholesteryl ester transfer protein activity assay kit
Select a Size
About This Item
Recommended Products
usage
sufficient for 100 fluorometric tests
application(s)
pharmaceutical
detection method
fluorometric
relevant disease(s)
cardiovascular diseases
storage temp.
2-8°C
Gene Information
human ... CETP(1071)
General description
Application
Features and Benefits
Suitability
Principle
related product
Storage Class Code
10 - Combustible liquids
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
-
What type of regression fitting is used for the product MAK106, and what is the method for calculating the pmoles shown in the typical standard curve in table 2?
1 answer-
Helpful?
-
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service