M9437
Greiner CELLSTAR® multiwell culture plates
48 wells (suspension culture with lid)
Synonym(s):
culture dishes, culture plates, multi dishes, Greiner CELLSTAR® multiwell culture plates
Select a Size
€307.00
Estimated to ship on31 May 2025
Select a Size
About This Item
polystyrene
€307.00
Estimated to ship on31 May 2025
Recommended Products
material
flat clear wells
polystyrene
sterility
sterile
feature
lid
packaging
pack of 1 ea (single packed)
manufacturer/tradename
Greiner 677102
size
48 wells
well volume
1.7 mL
well working volume
0.5- 1.0 mL
color
clear
binding type
Tissue Culture (TC)-treated surface
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
Features and Benefits
- Free of detectable DNase, RNase, human DNA Non-Cytotoxic
- Single position lids to preventcross-contamination
- High clarity and lowautofluorescence for clear observation of cell cultures
- Contains single position lids toprevent cross-contamination
- Lid enables optimal gas exchangewith minimal evaporation
- Alphanumeric well coding foreasy identification and tracking
- Seamless integration withautomated systems
- Convenient, easy-to-openpackaging
- Sterile and non-pyrogenic
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service