Select a Size
| Size/SKU | Availability | Price |
|---|---|---|
1 mg | Check Cart for Availability | € 1.610,00 € 1.207,50 |
About This Item
€ 1.207,50
List Price€ 1.610,00Save 25%Product Name
[Leu13]-Motilin porcine, ≥97% (HPLC)
Quality Segment
assay
≥97% (HPLC)
form
powder
color
white
UniProt accession no.
storage temp.
−20°C
SMILES string
CC[C@H](C)[C@H](NC(=O)[C@@H]1CCCN1C(=O)[C@@H](NC(=O)[C@@H](N)Cc2ccccc2)C(C)C)C(=O)N[C@@H](Cc3ccccc3)C(=O)N[C@@H]([C@@H](C)O)C(=O)N[C@@H](Cc4ccc(O)cc4)C(=O)NCC(=O)N[C@@H](CCC(O)=O)C(=O)N[C@@H](CC(C)C)C(=O)N[C@@H](CCC(N)=O)C(=O)N[C@@H](CCCNC(N)=N)C(=O)N[C@@H](CC(C)C)C(=O)N[C@@H](CCC(N)=O)C(=O)N[C@@H](CCC(O)=O)C(=O)N[C@@H](CCCCN)C(=O)N[C@@H](CCC(O)=O)C(=O)N[C@@H](CCCNC(N)=N)C(=O)N[C@@H](CC(N)=O)C(=O)N[C@@H](CCCCN)C(=O)NCC(=O)N[C@@H](CCC(N)=O)C(O)=O
InChI
1S/C121H190N34O35/c1-10-65(8)98(153-115(185)87-32-23-53-155(87)118(188)97(64(6)7)152-100(170)71(124)56-67-24-13-11-14-25-67)116(186)151-85(57-68-26-15-12-16-27-68)114(184)154-99(66(9)156)117(187)150-84(58-69-33-35-70(157)36-34-69)102(172)136-60-92(162)137-76(40-46-94(164)165)106(176)148-83(55-63(4)5)112(182)145-77(37-43-88(125)158)107(177)141-74(30-21-51-133-120(129)130)104(174)147-82(54-62(2)3)111(181)146-78(38-44-89(126)159)108(178)144-80(42-48-96(168)169)109(179)140-73(29-18-20-50-123)103(173)143-79(41-47-95(166)167)110(180)142-75(31-22-52-134-121(131)132)105(175)149-86(59-91(128)161)113(183)139-72(28-17-19-49-122)101(171)135-61-93(163)138-81(119(189)190)39-45-90(127)160/h11-16,24-27,33-36,62-66,71-87,97-99,156-157H,10,17-23,28-32,37-61,122-124H2,1-9H3,(H2,125,158)(H2,126,159)(H2,127,160)(H2,128,161)(H,135,171)(H,136,172)(H,137,162)(H,138,163)(H,139,183)(H,140,179)(H,141,177)(H,142,180)(H,143,173)(H,144,178)(H,145,182)(H,146,181)(H,147,174)(H,148,176)(H,149,175)(H,150,187)(H,151,186)(H,152,170)(H,153,185)(H,154,184)(H,164,165)(H,166,167)(H,168,169)(H,189,190)(H4,129,130,133)(H4,131,132,134)/t65-,66+,71-,72-,73-,74-,75-,76-,77-,78-,79-,80-,81-,82-,83-,84-,85-,86-,87-,97-,98-,99-/m0/s1
InChI key
GFOZQXMKARUPQI-HYOAWBISSA-N
Gene Information
pig ... MLN(397466)
Biochem/physiol Actions
Other Notes
1 of 1
This Item | |||
|---|---|---|---|
| form powder | form - | form powder | form powder |
| assay ≥97% (HPLC) | assay ≥97% (HPLC) | assay ≥97% (HPLC) | assay ≥90% (HPLC) |
| storage temp. −20°C | storage temp. −20°C | storage temp. −20°C | storage temp. −20°C |
| color white | color - | color - | color white |
| Quality Level 200 | Quality Level 200 | Quality Level 200 | Quality Level 200 |
| UniProt accession no. | UniProt accession no. | UniProt accession no. | UniProt accession no. - |
Storage Class
11 - Combustible Solids
wgk
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Gloves, type N95 (US)
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
![[Leu13]-Motilin porcine ≥97% (HPLC)](/deepweb/assets/sigmaaldrich/product/structures/276/894/aa9a75bb-198e-4685-976f-f7f2c5e55134/640/aa9a75bb-198e-4685-976f-f7f2c5e55134.png)


