Skip to Content
Merck
All Photos(1)

Key Documents

M4505

Sigma-Aldrich

Motilin porcine

≥97% (HPLC)

Sign Into View Organizational & Contract Pricing

Select a Size

0.1 MG
€167.00

€167.00


Estimated to ship on10 May 2025



Select a Size

Change View
0.1 MG
€167.00

About This Item

Empirical Formula (Hill Notation):
C120H188N34O35S
CAS Number:
Molecular Weight:
2699.05
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:
NACRES:
NA.32

€167.00


Estimated to ship on10 May 2025


Quality Level

Assay

≥97% (HPLC)

UniProt accession no.

storage temp.

−20°C

SMILES string

CC[C@H](C)[C@H](NC(=O)[C@@H]1CCCN1C(=O)[C@@H](NC(=O)[C@@H](N)Cc2ccccc2)C(C)C)C(=O)N[C@@H](Cc3ccccc3)C(=O)N[C@@H]([C@@H](C)O)C(=O)N[C@@H](Cc4ccc(O)cc4)C(=O)NCC(=O)N[C@@H](CCC(O)=O)C(=O)N[C@@H](CC(C)C)C(=O)N[C@@H](CCC(N)=O)C(=O)N[C@@H](CCCNC(N)=N)C(=O)N[C@@H](CCSC)C(=O)N[C@@H](CCC(N)=O)C(=O)N[C@@H](CCC(O)=O)C(=O)N[C@@H](CCCCN)C(=O)N[C@@H](CCC(O)=O)C(=O)N[C@@H](CCCNC(N)=N)C(=O)N[C@@H](CC(N)=O)C(=O)N[C@@H](CCCCN)C(=O)NCC(=O)N[C@@H](CCC(N)=O)C(O)=O

InChI

1S/C120H188N34O35S/c1-9-64(6)97(152-114(184)86-31-22-53-154(86)117(187)96(63(4)5)151-99(169)70(123)56-66-23-12-10-13-24-66)115(185)150-84(57-67-25-14-11-15-26-67)113(183)153-98(65(7)155)116(186)149-83(58-68-32-34-69(156)35-33-68)101(171)135-60-91(161)136-75(39-45-93(163)164)105(175)147-82(55-62(2)3)111(181)145-77(37-43-88(125)158)106(176)140-73(29-20-51-132-119(128)129)103(173)146-80(48-54-190-8)110(180)142-76(36-42-87(124)157)107(177)144-79(41-47-95(167)168)108(178)139-72(28-17-19-50-122)102(172)143-78(40-46-94(165)166)109(179)141-74(30-21-52-133-120(130)131)104(174)148-85(59-90(127)160)112(182)138-71(27-16-18-49-121)100(170)134-61-92(162)137-81(118(188)189)38-44-89(126)159/h10-15,23-26,32-35,62-65,70-86,96-98,155-156H,9,16-22,27-31,36-61,121-123H2,1-8H3,(H2,124,157)(H2,125,158)(H2,126,159)(H2,127,160)(H,134,170)(H,135,171)(H,136,161)(H,137,162)(H,138,182)(H,139,178)(H,140,176)(H,141,179)(H,142,180)(H,143,172)(H,144,177)(H,145,181)(H,146,173)(H,147,175)(H,148,174)(H,149,186)(H,150,185)(H,151,169)(H,152,184)(H,153,183)(H,163,164)(H,165,166)(H,167,168)(H,188,189)(H4,128,129,132)(H4,130,131,133)/t64-,65+,70-,71-,72-,73-,74-,75-,76-,77-,78-,79-,80-,81-,82-,83-,84-,85-,86-,96-,97-,98-/m0/s1

InChI key

LVRVABPNVHYXRT-BQWXUCBYSA-N

Gene Information

pig ... MLN(397466)

Looking for similar products? Visit Product Comparison Guide

Amino Acid Sequence

Phe-Val-Pro-Ile-Phe-Thr-Tyr-Gly-Glu-Leu-Gln-Arg-Met-Gln-Glu-Lys-Glu-Arg-Asn-Lys-Gly-Gln

Other Notes

Lyophilized from 0.1% TFA in H2O

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Central, but not peripheral application of motilin increases c-Fos expression in hypothalamic nuclei in the rat brain
Mei Wu
Histochemistry and Cell Biology (2005)
Chen Shaodong et al.
Lipids in health and disease, 12, 21-21 (2013-02-22)
To investigate the influence and mechanism of combining exercise with diet control on a model of lipid metabolism rat induced by high fat diet. Twenty-four male Wistar rats were randomly divided into 3 groups of 8: normal, model and intervention.
Li Chen et al.
Zhen ci yan jiu = Acupuncture research, 37(3), 229-232 (2012-09-01)
To observe the curative effect of autologous blood point injection on diabetic gastroparesis and to study its underlying mechanism. A total of 42 type-II diabetes outpatients and inpatients were randomly and equally divided into treatment group and control group according
Rudolf L Riepl et al.
PloS one, 7(9), e44445-e44445 (2012-09-13)
Acute mountain sickness (AMS) is characterized by headache often accompanied by gastrointestinal complaints that vary from anorexia through nausea to vomiting. The aim of this study was to investigate the influence of high altitude on plasma levels of gastroenteropancreatic (GEP)
Jing He et al.
Journal of molecular evolution, 75(3-4), 112-118 (2012-11-10)
The motilin receptor gene was lost in the ancestral lineage of rodents. Subsequently, the gene encoding its ligand, motilin, has experienced different evolutionary fates. Previous genomic analyses had shown that the motilin gene (MLN) became a pseudogene independently in the

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service