M2625
DL-Mandelic acid
Synonym(s):
(±)-α-Hydroxyphenylacetic acid
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Linear Formula:
C6H5CH(OH)COOH
CAS Number:
Molecular Weight:
152.15
Beilstein:
510011
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
Recommended Products
SMILES string
OC(C(O)=O)c1ccccc1
InChI
1S/C8H8O3/c9-7(8(10)11)6-4-2-1-3-5-6/h1-5,7,9H,(H,10,11)
InChI key
IWYDHOAUDWTVEP-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Dam. 1
Storage Class Code
13 - Non Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
P Vodicka et al.
Carcinogenesis, 16(7), 1473-1481 (1995-07-01)
Occupational exposure to styrene was studied in nine workers of a hand lamination plant in Bohemia. Personal dosimeters were used to monitor the styrene workplace exposure, and the levels of styrene in blood and mandelic acid in urine were measured.
Ludmila Krivánková et al.
Electrophoresis, 24(3), 505-517 (2003-02-06)
Effects originating from the variability of the sample matrix can be efficiently eliminated when the separation conditions are selected so that compounds of like charge with high concentration referred to as macrocomponents are embodied into the system of transient isotachophoresis.
A Kimura et al.
Pediatric research, 45(4 Pt 1), 603-609 (1999-04-15)
Unusual bile acids, such as unsaturated ketonic and 7beta-hydroxylated bile acids, have been detected in urine early in life. To elucidate the normal profiles of usual and unusual urinary bile acids in the neonatal and pediatric periods, we measured the
David A Ostrov et al.
Antimicrobial agents and chemotherapy, 51(10), 3688-3698 (2007-08-08)
The bacterial type II topoisomerases DNA gyrase and topoisomerase IV are validated targets for clinically useful quinolone antimicrobial drugs. A significant limitation to widely utilized quinolone inhibitors is the emergence of drug-resistant bacteria due to an altered DNA gyrase. To
H Crespi et al.
The Journal of investigative dermatology, 78(6), 493-497 (1982-06-01)
The purpose of this study was to evaluate the in vivo state of both branches of the autonomic nervous system in children with chronic atopic dermatitis. In 15 patients, age 4 to 11, the following parameters were analyzed: (1) basal
Chromatograms
suitable for GCapplication for HPLCsuitable for GCOur team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service