8.06913
DL-Mandelic acid
for synthesis
Synonym(s):
DL-Mandelic acid, DL-α-Hydroxyphenylacetic acid, Phenylglycollic acid
Select a Size
Select a Size
About This Item
Recommended Products
vapor pressure
0.01 Pa ( 50 °C)
Quality Level
Assay
≥99.0% (acidimetric)
form
crystals
pH
2.3 (10 g/L in H2O)
mp
119-121 °C
solubility
139 g/L
density
1.31 g/cm3 at 20.2 °C
bulk density
750 kg/m3
storage temp.
2-30°C
SMILES string
OC(c1ccccc1)C(=O)O
InChI
1S/C8H8O3/c9-7(8(10)11)6-4-2-1-3-5-6/h1-5,7,9H,(H,10,11)
InChI key
IWYDHOAUDWTVEP-UHFFFAOYSA-N
Related Categories
Application
- Kinetics and Mechanism of Oxidation of Phenyl Acetic Acid and Dl-mandelic Acid by Permanganate in Acid Medium: This study explores the oxidation mechanisms of DL-Mandelic acid and its comparison with phenyl acetic acid, highlighting its higher reactivity. (KV Ratnam et al., 2014).
- DL-mandelic acid intercalated Zn-Al layered double hydroxide: Investigates a novel antimicrobial layered material incorporating DL-Mandelic acid, with potential applications in healthcare products. (LP Tang et al., 2018).
Analysis Note
Melting range (lower value): ≥ 116 °C
Melting range (upper value): ≤ 121 °C
Identity (IR): conforms
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Dam. 1
Storage Class Code
13 - Non Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service