Skip to Content
Merck
All Photos(1)

Key Documents

L9773

Sigma-Aldrich

Luteinizing Hormone from equine pituitary

pkg of ~15 units/vial

Synonym(s):

ICSH, LH, PLH

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
EC Number:
MDL number:
UNSPSC Code:
51111800

form

powder

Quality Level

packaging

pkg of ~15 units/vial

solubility

water: 50 mg/mL, clear to slightly hazy, colorless to faintly yellow

UniProt accession no.

shipped in

ambient

storage temp.

2-8°C

Gene Information

Application

Luteinizing hormone (LH) has been used to study effect of LH on the development of preantral follicles for nuclear maturation of ovine oocytes and as an endocrine signal to study circadian phase in the ovary.

Biochem/physiol Actions

Two chain glycoprotein hormone, the α-chain is not active, biological specificity is attributed to the β-chain; induces ovulation, spermatogenesis, and synthesis of sex steroids.

Packaging

Package size based on NIH standard LH-S1 using the rat Leydig cell assay

Quantity

One NIH LH-S1 unit = approx. 2,000 I.U.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Repr. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Timing of the Ovarian Circadian Clock Is Regulated by Gonadotropins
Tomoko Y
Endocrinology (2009)
Nuclear maturation of ovine oocytes in cultured preantral follicles
G Tamilmani
The Journal of Small Animal Practice (2005)
Tomoko Yoshikawa et al.
Endocrinology, 150(9), 4338-4347 (2009-06-13)
The timing of ovulation is critically important to the success of reproduction. Current thinking attributes the timing of ovulation to LH secretion by the pituitary, itself timed by signals from the hypothalamus. The discovery of an internal circadian timer in
Rachel Y Cheong et al.
Endocrinology, 155(4), 1418-1427 (2014-01-31)
The negative feedback mechanism through which 17β-estradiol (E2) acts to suppress the activity of the GnRH neurons remains unclear. Using inducible and cell-specific genetic mouse models, we examined the estrogen receptor (ER) isoforms expressed by neurons that mediate acute estrogen
Hidetoshi Hasuwa et al.
Science (New York, N.Y.), 341(6141), 71-73 (2013-06-15)
Ovulation in the mouse and other mammals is controlled by hormones secreted by the hypothalamo-pituitary-ovarian axis. We describe anovulation and infertility in female mice lacking the microRNAs miR-200b and miR-429. Both miRNAs are strongly expressed in the pituitary gland, where

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service