Skip to Content
Merck
All Photos(1)

Key Documents

L6385

Sigma-Aldrich

α-Lactalbumin from bovine milk

For use as a marker in SDS-PAGE

Synonym(s):

alpha-lactalbumin

Sign Into View Organizational & Contract Pricing

Select a Size

1 VIAL
€56.00

€56.00


Estimated to ship on29 May 2025



Select a Size

Change View
1 VIAL
€56.00

About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352202
NACRES:
NA.61

€56.00


Estimated to ship on29 May 2025


biological source

bovine milk

Quality Level

form

powder

mol wt

~14.2 kDa

packaging

vial of 5 mg

concentration

>5 mg per vial protein (biuret)

technique(s)

electrophoresis: suitable

solubility

H2O: soluble 10 mg/mL

UniProt accession no.

storage temp.

2-8°C

Gene Information

Looking for similar products? Visit Product Comparison Guide

General description

α-Lactalbumin is a small, globular, whey protein that has been found in all milk studied to date. It is a metalloprotein of approximately 14 kDa produced in the mammary glands.[1]

Application

α-Lactalbumin from bovine milk is suitable for use:
  • as an electrophoresis marker, with a molar mass of approximately 14,200Da[2][3][4]
  • in a study to investigate selective binding of proteins on charged surface iron oxide nanoparticles via reverse charge parity model[5]
α-Lactalbumin was used in the reduction of the antigenicity of whey proteins by lactic acid fermentation.[6]

Biochem/physiol Actions

α-Lactalbumin consists of a single polypeptide chain with 8 cysteines which form disulfide bridges. α-Lactalbumin binds several metal ions, including calcium, which is thought to play a role in the regeneration of native α-lactalbumin from the reduced, denatured form.[7] α-Lactalbumin also has a distinct zinc binding site that is thought to play a role in the binding of the lactose synthase complex. [8][9] The mature protein consists of 123 amino acid residues (14 kD), and it has a three-dimensional structure with 1.7 Angstrom resolution, demonstrating four a-helices and a triple stranded antiparallel β-sheet.[1]
Alters the substrate specificity of galactosyltransferase to increase the rate of lactose formation; the complex of galactosyltransferase and α-lactalbumin is called lactose synthase.
Alters the substrate specificity of galactosyltransferase to increase the rate of lactose formation; the complex of galactosyltransferase and α-lactalbumin is called lactose synthase. Site-directed mutagenesis of Asp87 or Asp88 to Ala completely abolishes the strong calcium binding affinity and reduces the stimulation of lactose synthase to <3.5% of the maximal rate.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Haidong Tan et al.
Biotechnology letters, 27(16), 1177-1182 (2005-09-15)
Recombinant human interleukin-1 receptor antagonist (rHuIL-1ra) was produced in E. coli as an inclusion body. rHuIL-1ra was purified to Over 98% purity by anion exchange chromatography after on-column refolding. The optimized processes produced more than 2 g pure refolded rHuIL-1ra
S Yefimov et al.
Journal of biochemical and biophysical methods, 42(1-2), 65-78 (2000-01-27)
Five SDS-proteins, ranging in molecular weight from 14 to 66 kDa, were detected without covalent fluorescent labeling by the automated gel electrophoresis apparatus with intermittent fluorescence scanning (HPGE apparatus, LabIntelligence) during electrophoresis in barbiturate buffer in the presence of Cascade
Eric Lorent et al.
Vaccine, 26(3), 399-410 (2007-12-14)
The structure of the ectodomain of the hepatitis C envelope glycoprotein E1 (E1s) was characterised by spectroscopic methods. Monomeric E1s was purified from a mammalian and from a Hansenula polymorpha cell lysate, and cysteine-blocked monomers were reconstituted into stable particles.
Selective binding of proteins on functional nanoparticles via reverse charge parity model: an in vitro study.
Ghosh G, et, al.
Materials Research Express, 1 (2014)
Guanhao Bu et al.
Journal of the science of food and agriculture, 90(12), 2015-2020 (2010-06-29)
The main whey proteins alpha-lactalbumin (alpha-LA) and beta-lactoglobulin (beta-LG) are considered as the major allergens in cow's milk. Microbial fermentation can produce some proteolytic enzymes, which can induce the degradation of milk protein allergens. In this study, the effects of

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service