Skip to Content
Merck
All Photos(1)

Key Documents

L-046

Supelco

β-Lactoglobulin from bovine milk

certified reference material, ampule of 100 mg, Cerilliant®

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352202
NACRES:
NA.24
Pricing and availability is not currently available.

grade

certified reference material

Quality Level

form

solid

feature

Snap-N-Spike®/Snap-N-Shoot®

packaging

ampule of 100 mg

manufacturer/tradename

Cerilliant®

technique(s)

liquid chromatography (LC): suitable

application(s)

food and beverages

format

neat

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

General description

β-Lactoglobulin A & B is the major whey protein of cow and sheep′s milk and is also present in many other mammalian species with an interesting exception being humans. It is considered a food allergen. This full-length protein CRM is suitable as starting material for use in calibrators or controls for a variety of LC-MS/MS testing applications including allergen testing, infant formula testing and dietary or nutritional testing applications.

Application


  • beta-Lactoglobulin protein purification: Utilized in developing high-capacity anion exchangers, beta-Lactoglobulin aids in protein purification processes essential for pharmaceutical research and production, improving efficiency and scale-up capabilities (Ng and Snyder, 2013).

Features and Benefits

  • Multiple analytical techniques have been employed to identify and comprehensively characterize the material.
  • The certified property value is traceable to national institute of standards and technology (NIST) through calibration verification using NIST traceable weights.
  • Balances and NIST traceable weights are calibrated.
  • Long term stability has been evaluated for freezer storage (-10 °C to -25 °C) conditions.

Preparation Note

  • Weigh the material before usage.
  • The amount to be weighed should be determined based on the certified property value. Please note that the material is hygroscopic.
  • To maintain accuracy, minimize exposure to moisture during the weighing process.
  • For quantitative applications, the minimum sample size for intended use is 20 mg.
  • If a smaller sample size is necessary, it is advisable to increase the uncertainty by a factor of 2 for accurate results.

Other Notes

For analytical and R&D purposes only. This product′s purpose is for testing. THIS PRODUCT IS NOT INTENDED FOR HUMAN OR THERAPEUTIC USE.

Legal Information

CERILLIANT is a registered trademark of Merck KGaA, Darmstadt, Germany
Snap-N-Shoot is a registered trademark of Cerilliant Corporation
Snap-N-Spike is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

CRMs of milk proteins aid accurate testing for infant formula, ensuring compliance and quality control.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service