Skip to Content
Merck

Skip To

438192

Anti-LRP, Light Chain Mouse mAb (5A6)

liquid, clone 5A6, Calbiochem®

Synonym(s):

Anti-Low Density Lipoprotein Receptor-Related Protein

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
100 μg
Check Cart for Availability
€ 428,00

About This Item

NACRES:
NA.43
UNSPSC Code:
12352203
Clone:
5A6, monoclonal
Species reactivity:
rabbit, mouse, human
Application:
Citations:
7

€ 428,00


Check Cart for Availability

Request a Custom Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

mouse

Quality Segment

antibody form

purified antibody

antibody product type

primary antibodies

clone

5A6, monoclonal

form

liquid

does not contain

preservative

species reactivity

rabbit, mouse, human

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze, avoid repeated freeze/thaw cycles

dilution

(Frozen Sections (1-5 µg/mL)
Immunoblotting (1-5 µg/mL)
Paraffin Sections (1-5 µg/mL, )

isotype

IgG2

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... LRP1(4035)

General description

Purified mouse monoclonal antibody. Recognizes the ~85 kDa LRP light chain protein under non-reducing conditions.
Recognizes the ~85 kDa LRP protein.
This Anti-LRP, Light Chain Mouse mAb (5A6) is validated for use in Frozen Sections, Immunoblotting, Paraffin Sections for the detection of LRP, Light Chain.

Immunogen

Human
purified, human placenta LRP

Application

Frozen Sections (1-5 µg/ml)

Immunoblotting (1-5 µg/ml)

Paraffin Sections (1-5 µg/ml, see application references)

Packaging

Please refer to vial label for lot-specific concentration.

Physical form

In 100 mM NaCl, 50 mM sodium phosphate, 1 mM EDTA, pH 6.6.

Preparation Note

Following initial thaw, aliquot and freeze (-20°C).

Other Notes

Mikhailenko, I., et al. 2001. J. Biol. Chem.276, 39484.
Rebeck, W.G., et al. 2001. Mol. Brain Res.87, 238.
Coukos, G., et al. 1994. Am. J. Pathol.144, 383.
Daugherty, A., and Rateri, D.L. 1994. Arterioscler Thromb.14, 2017.
Strickland, D.K., et al. 1990. J. Biol. Chem.265, 17401.
Variables associated with assay conditions will dictate the proper working dilution.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Toxicity: Standard Handling (A)

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
ST1685AP1145OP145
species reactivity

rabbit, mouse, human

species reactivity

human

species reactivity

human

species reactivity

human

Gene Information

human ... LRP1(4035)

Gene Information

human ... TRPA1(8989)

Gene Information

human ... IL13RA2(3598)

Gene Information

human ... MDM2(4193)

antibody form

purified antibody

antibody form

purified antibody

antibody form

purified antibody

antibody form

purified antibody

biological source

mouse

biological source

mouse

biological source

mouse

biological source

mouse

clone

5A6, monoclonal

clone

6G8, monoclonal

clone

2E10, monoclonal

clone

4B2C1.11, monoclonal

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C


Still not finding the right product?

Try our Product Selector Tool to narrow your options


Storage Class

10 - Combustible liquids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters