HS120498
Tube rack with rotating shelf, for 5/15 mL or 16 mm tubes
to hold, 20 x 10-17 mm tubes, purple rack, natural rack
All Photos(1)
About This Item
Recommended Products
material
natural rack
polypropylene
purple rack
feature
to hold 20 x 10-17 mm tubes
packaging
pack of 1 ea
manufacturer/tradename
Heathrow Scientific 120498
L × W × H
309 mm × 107 mm × 77 mm
General description
Rotate with ease to accommodate your needs
With bench top space at a premium, it′s important for research and clinical labs working across a range of applications to efficiently use the space available. Unlike other tube racks, these multi-purpose racks are specifically designed with the flexibility to hold multiple tube sizes firmly in place to assist technicians constantly alternating between sample vessels.
The rack incorporates two moving hinged shelves that can be rotated to an up or down position to accommodate the size of the tube being used. When working with samples, the shelves can be rotated to an angle position to facilitate more comfortable pipetting and allow better visibility. The tube rack wells have a unique teardrop design that enables easier insertion and better retention of tubes over traditional designs.
The 5 mL/15 mL/16mm Tube Rack rotates with ease to accommodate your needs. This multi-purpose rack is designed with the flexibility to hold 20 - 5 mL, 20 - 15 mL or 16mm tubes OR to hold 40 - 5 mL, 40 - 15 mL or 16 mm tubes. The rack incorporates two moving hinged upper shelves that can be rotated to an up or down position to accommodate the size of the tube being used. In addition, the rack can also hold clinical tubes of a similar diameter (16 mm) and height.
Additional Features:
With bench top space at a premium, it′s important for research and clinical labs working across a range of applications to efficiently use the space available. Unlike other tube racks, these multi-purpose racks are specifically designed with the flexibility to hold multiple tube sizes firmly in place to assist technicians constantly alternating between sample vessels.
The rack incorporates two moving hinged shelves that can be rotated to an up or down position to accommodate the size of the tube being used. When working with samples, the shelves can be rotated to an angle position to facilitate more comfortable pipetting and allow better visibility. The tube rack wells have a unique teardrop design that enables easier insertion and better retention of tubes over traditional designs.
The 5 mL/15 mL/16mm Tube Rack rotates with ease to accommodate your needs. This multi-purpose rack is designed with the flexibility to hold 20 - 5 mL, 20 - 15 mL or 16mm tubes OR to hold 40 - 5 mL, 40 - 15 mL or 16 mm tubes. The rack incorporates two moving hinged upper shelves that can be rotated to an up or down position to accommodate the size of the tube being used. In addition, the rack can also hold clinical tubes of a similar diameter (16 mm) and height.
Additional Features:
- Ergonomic handles for ease of transporting the rack
- Molded grid on both sides of the shelves for quick sample identification and retrieval
- Autoclavable
- US Patent Pending
The Multi-tube rack is designed to hold 5, 10, 15 mL tubes (or any tube between 10-17 mm in diameter). The rack design incorporates a total of four moving hinged upper shelves that can be positioned for the size of the tube being used. It is important for research and clinical labsworking across a range of applications to efficiently use the space available. Unlike other tube racks, these multi-purpose racks are specifically designed with the flexibility to hold multiple tube sizes firmly in place to assist technicians constantlyalternating between sample vessels.
Features and Benefits
- Rack holds tubes with a diameter of 10-17 mm by using the different shelves
- Unique tear drop shaped openings (top tray) for better retention of tubes
- Position numbering on both sides of the upper shelves
- Shelves easily rotate to accommodate the different size tubes
- Autoclavable
- Ergonomic handles for ease of use