Skip to Content
Merck
All Photos(7)

Key Documents

HPA026506

Sigma-Aldrich

Anti-DARS2 antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution, Ab1

Synonym(s):

Anti-AspRS, Anti-Aspartate--tRNA ligase, Anti-Aspartyl-tRNA synthetase, mitochondrial

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
€544.00

€544.00


Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)


Select a Size

Change View
100 μL
€544.00

About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

€544.00


Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

independent
Learn more about Antibody Enhanced Validation

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunohistochemistry: 1:20-1:50

immunogen sequence

EVTLCGWIQYRRQNTFLVLRDFDGLVQVIIPQDESAASVKKILCEAPVESVVQVSGTVISRPAGQENPKMPTGEIEIKVKTAELLNACKKLPFEIKNFVKK

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... DARS2(55157)

General description

Aspartyl-tRNA synthetase 2, mitochondrial (DARS2) is a homodimeric enzyme. The gene encoding it is localized on human chromosome 1q25.1.

Immunogen

Aspartyl-tRNA synthetase, mitochondrial Precursor recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

Aspartyl-tRNA synthetase 2, mitochondrial (DARS2) has a crucial role in the attachment of aspartate to the right mitochondrial transfer RNA. Mutations in the gene encoding it have been associated with leukoencephalopathy with brainstem and spinal cord involvement and elevated lactate (LBSL).

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST76658

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A novel homozygous mutation of DARS2 may cause a severe LBSL variant.
Miyake N
Clinical Genetics, 80(3), 293-296 (2011)
Pathogenic mutations causing LBSL affect mitochondrial aspartyl-tRNA synthetase in diverse ways.
van Berge L
The Biochemical Journal, 450(2), 345-350 (2013)
DARS-associated leukoencephalopathy can mimic a steroid-responsive neuroinflammatory disorder.
Wolf NI, et.l
Neurology, 84(3), 226-230 (2015)
Laura van Berge et al.
Brain : a journal of neurology, 137(Pt 4), 1019-1029 (2014-02-26)
Leukoencephalopathy with brainstem and spinal cord involvement and lactate elevation is a disorder caused by recessive mutations in the gene DARS2, which encodes mitochondrial aspartyl-tRNA synthetase. Recent observations indicate that the phenotypic range of the disease is much wider than

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service