Skip to Content
Merck

Skip To

ABE198

Anti-Sirt5 Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

NAD-dependent deacetylase sirtuin-5, SIR2-like protein 5

Sign In to View Organizational & Contract Pricing.

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
eCl@ss:
32160702
Conjugate:
unconjugated
Clone:
polyclonal
Application:
IHC, WB
Citations:
4


biological source

rabbit

Quality Segment

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

rat, human

species reactivity (predicted by homology)

chimpanzee (based on 100% sequence homology), rhesus macaque (based on 100% sequence homology)

technique(s)

immunohistochemistry: suitable, western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... SIRT5(23408)

General description

Sirtuin (SIRT5) is known as a nicotinamide adenine dinucleotide- dependent deacetylase that is a member of the silent information regulator 2 (Sir2) family of proteins that is important in aging, metabolism and disease. SIRT5 is expressed in a variety of tissues. Two different isoform have been described. Isoform 1 is reported to be express in the mitochondria and cytoplasm while isoform 2 appears to be localized to the mitochondria only. SIRT5 has also been linked to many primate-specific functions. SIRT5 has recently been implicated as having a role in adaptation to fasting, caloric restriction and high protein diets.
~34 kDa observed. Two isoforms at ~33 and ~34 kDa are known to exist.

Immunogen

KLH-conjugated linear peptide corresponding to human Sirt5.

Application

Immunohistochemistry Analysis: 1:500 dilution of a representative lot of this antibody detected Sirt5 in convoluted tubules of normal rat kidney tissue.
Use Anti-Sirt5 Antibody (Rabbit Polyclonal Antibody) validated in WB, IHC to detect Sirt5 also known as NAD-dependent deacetylase sirtuin-5, SIR2-like protein 5.

Analysis Note

Evaluated by Western Blot in HeLa cell lysate.

Western Blot Analysis: 0.5 µg/mL of this antibody detected Sirt5 on 10 µg of HeLa cell lysate.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
AV32390AV40125SAB3500496
antibody form

affinity isolated antibody

antibody form

affinity isolated antibody

antibody form

IgG fraction of antiserum

antibody form

affinity isolated antibody

biological source

rabbit

biological source

rabbit

biological source

rabbit

biological source

chicken

conjugate

unconjugated

conjugate

unconjugated

conjugate

unconjugated

conjugate

unconjugated

Quality Level

100

Quality Level

100

Quality Level

-

Quality Level

100

Gene Information

human ... SIRT5(23408)

Gene Information

human ... SIRT5(23408)

Gene Information

human ... SIRT5(23408)

Gene Information

human ... SIRT5(23408)

species reactivity

rat, human

species reactivity

goat, guinea pig, human

species reactivity

human, goat

species reactivity

human, mouse


Still not finding the right product?

Try our Product Selector Tool to narrow your options


Storage Class

12 - Non Combustible Liquids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters