Skip to Content
Merck
All Photos(3)

Key Documents

HPA024679

Sigma-Aldrich

Anti-TONSL antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-I-kappa-B-related protein, Anti-IkappaBR, Anti-Inhibitor of kappa B-related protein, Anti-NF-kappa-B inhibitor-like protein 2, Anti-NFKBIL2, Anti-Nuclear factor of kappa light polypeptide gene enhancer in B-cells inhibitor-like 2

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
€544.00

€544.00


Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)


Select a Size

Change View
100 μL
€544.00

About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

€544.00


Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:50-1:200

immunogen sequence

WNRRNDMGETLLHRACIEGQLRRVQDLVRQGHPLNPRDYCGWTPLHEACNYGHLEIVRFLLDHGAAVDDPGGQGCEGITPLHDALNCGHFEVAELLLERGASVTLRTRK

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... NFKBIL2(4796)

General description

The gene TONSL (tonsoku-like) is mapped to human chromosome 8q24.3. The protein contains three ankyrin-repeat motifs.

Immunogen

NF-kappa-B inhibitor-like protein 2 recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies®Powered by Atlas Antibodies is developed and validated by the Human Protein Atlas (HPA) project . Each antibody is tested by immunohistochemistry against hundreds of normal and disease tissues. These images can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.
Anti-TONSL antibody produced in rabbit has been used in Western blotting.[1]
Applications in which this antibody has been used successfully, and the associated peer-reviewed papers, are given below.
Western Blotting (1 paper)

Biochem/physiol Actions

TONSL (tonsoku-like) associates with NF-κB (nuclear factor κ B). Mutations in TONSL increases the risk to invasive pneumococcal disease. MMS22L (MMS22 like)-TONSL complex provides genomic stability at the time of DNA replication. Mutations in ankyrin-repeat domains (present in the TONSL protein) negatively affects genome stability and cell viability.[1]

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST70954

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

D A Norman et al.
Annals of human genetics, 64(Pt 1), 15-23 (2001-03-14)
The inhibitors of NF-kappaB (IkappaBs) play an important role in the regulation of the NF-kappaB pathway. IkappaBR (for IkappaB-Related) is proposed to be a novel member of this family. We report the cloning and characterization of the region of the
Stephen J Chapman et al.
Critical care (London, England), 14(6), R227-R227 (2010-12-22)
Streptococcus pneumoniae remains a major global health problem and a leading cause of death in children worldwide. The factors that influence development of pneumococcal sepsis remain poorly understood, although increasing evidence points towards a role for genetic variation in the
Wojciech Piwko et al.
The EMBO journal, 29(24), 4210-4222 (2010-11-30)
Cullin 4 (Cul4)-based ubiquitin ligases emerged as critical regulators of DNA replication and repair. Over 50 Cul4-specific adaptors (DNA damage-binding 1 (Ddb1)-Cul4-associated factors; DCAFs) have been identified and are thought to assemble functionally distinct Cul4 complexes. Using a live-cell imaging-based
Giulia Saredi et al.
Nature, 534(7609), 714-718 (2016-06-25)
After DNA replication, chromosomal processes including DNA repair and transcription take place in the context of sister chromatids. While cell cycle regulation can guide these processes globally, mechanisms to distinguish pre- and post-replicative states locally remain unknown. Here we reveal

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service