Skip to Content
Merck
All Photos(6)

Key Documents

HPA023086

Sigma-Aldrich

Anti-SGPL1 antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution, Ab2

Synonym(s):

Anti-SP-lyase, Anti-Sphingosine-1-phosphate aldolase, Anti-Sphingosine-1-phosphate lyase 1, Anti-hSPL

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
€544.00

€544.00


Check Cart for Availability


Select a Size

Change View
100 μL
€544.00

About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

€544.00


Check Cart for Availability

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

availability

not available in USA

enhanced validation

independent
Learn more about Antibody Enhanced Validation

technique(s)

immunohistochemistry: 1:500- 1:1000

immunogen sequence

GEEKLTELLVKAYGDFAWSNPLHPDIFPGLRKIEAEIVRIACSLFNGGPDSCGCVTSGGTESILMACKAYRDLAFEKGIKTPEIVAPQSAHAAFNKAASYFGMKIVRVPLTKMMEVDVRAMRRAISRNTAMLVCSTPQFPHGVI

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... SGPL1(8879)

General description

SGPL1 (sphingosine-1-phosphate lyase 1) is strongly expressed in differentiated enterocytes and is involved in catabolism of S1P (sphingosine-1-phosphate). The gene is mapped to human chromosome 10q21.

Immunogen

Sphingosine-1-phosphate lyase 1 recombinant protein epitope signature tag (PrEST)

Application

Anti-SGPL1 antibody produced in rabbit, a Prestige Antibody, is developed and validated by the Human Protein Atlas (HPA) project . Each antibody is tested by immunohistochemistry against hundreds of normal and disease tissues. These images can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. The antibodies are also tested using immunofluorescence and western blotting. To view these protocols and other useful information about Prestige Antibodies and the HPA, visit sigma.com/prestige.
Applications in which this antibody has been used successfully, and the associated peer-reviewed papers, are given below.
Immunohistochemistry (1 paper)
Western Blotting (1 paper)

Biochem/physiol Actions

SGPL1 (sphingosine-1-phosphate lyase 1) converts S1P (sphingosine-1-phosphate) into hexadecenal and ethanolamine phosphate. It is also activated in the presence of DNA damage. Upregulation of SGPL1 enhances apoptosis.[1]

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST74229

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sandra Colié et al.
Cancer research, 69(24), 9346-9353 (2009-11-26)
Sphingosine 1-phosphate (S1P) is a bioactive sphingolipid metabolite involved in cancer development through stimulation of cell survival, proliferation, migration, and angiogenesis. Irreversible degradation of S1P is catalyzed by S1P lyase (SPL). The human SGPL1 gene that encodes SPL maps to
A Kumar et al.
Cell death & disease, 2, e119-e119 (2011-03-04)
The injurious consequences of ionizing radiation (IR) to normal human cells and the acquired radioresistance of cancer cells represent limitations to cancer radiotherapy. IR induces DNA damage response pathways that orchestrate cell cycle arrest, DNA repair or apoptosis such that
Mariam Gachechiladze et al.
Oncology letters, 18(2), 1259-1266 (2019-08-20)
Sphingosine 1-phosphate (S1P) is a bioactive lipid metabolite associated with cancer cell proliferation, survival, migration and regulation of tumor angiogenesis in various cellular and animal models. Sphingosine kinase-1 (SphK1) and S1P lyase are the main enzymes that respectively control the
Johnatan Ceccom et al.
Acta neuropathologica communications, 2, 12-12 (2014-01-29)
The accumulation of beta amyloid (Aβ) peptides, a hallmark of Alzheimer's disease (AD) is related to mechanisms leading to neurodegeneration. Among its pleiotropic cellular effects, Aβ accumulation has been associated with a deregulation of sphingolipid metabolism. Sphingosine 1-phosphate (S1P) derived
Emilie Degagné et al.
The Journal of clinical investigation, 124(12), 5368-5384 (2014-10-28)
Growing evidence supports a link between inflammation and cancer; however, mediators of the transition between inflammation and carcinogenesis remain incompletely understood. Sphingosine-1-phosphate (S1P) lyase (SPL) irreversibly degrades the bioactive sphingolipid S1P and is highly expressed in enterocytes but downregulated in

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service