Skip to Content
Merck
All Photos(8)

Key Documents

HPA018246

Sigma-Aldrich

Anti-FXR1 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-Fragile X mental retardation syndrome-related protein 1, Anti-hFXR1p

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
€505.00

€505.00


Check Cart for Availability


Select a Size

Change View
100 μL
€505.00

About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.43

€505.00


Check Cart for Availability

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

rat, mouse, human

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:200-1:500

immunogen sequence

TESERKDELSDWSLAGEDDRDSRHQRDSRRRPGGRGRSVSGGRGRGGPRGGKSSISSVL

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... FXR1(8087)

General description

The gene FXR1 (fragile X mental retardation syndrome-related protein 1) is mapped to human chromosome 3q28. FXR1 belongs to fragile X related family. It is an autosomal paralog of fragile X mental retardation 1. FXR1 is a RNA binding protein and is localized in the cytoplasm. RT-PCR analysis showed FXR1 expression in brain, heart, kidney and testis.

Immunogen

Fragile X mental retardation syndrome-related protein 1 recombinant protein epitope signature tag (PrEST)

Application

Anti-FXR1 antibody produced in rabbit, a Prestige Antibody, is developed and validated by the Human Protein Atlas (HPA) project . Each antibody is tested by immunohistochemistry against hundreds of normal and disease tissues. These images can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. The antibodies are also tested using immunofluorescence and western blotting.[1] To view these protocols and other useful information about Prestige Antibodies and the HPA, visit sigma.com/prestige.

Biochem/physiol Actions

Fragile X mental retardation syndrome-related protein 1 (FXR1) plays an important role in development as FXR1 knockout mice die early during embryogenesis. FXR1 is important for proper development of the muscle tissue. Facioscapulohumeral muscular dystrophy myoblasts show abnormal expression of FXR1 which deregulates the proper metabolism of muscle specific mRNAs. Similarly, miR369-3 mediated association of argonaute 2 and FXR1 with AU-rich elements directs translational activation. FXR1 forms a complex with oncogenes, protein kinase C, iota and epithelial cell transforming-2, resulting in tumor progression and non-small cell lung cancer growth. Binding of plakophilins 1/3 with FXR1 is important for stability of specific mRNAs.[1] FXR1 also interacts with tudor domain-containing protein-3 and Bcl-2-associated transcription factor-1. Glycogen synthase kinase 3β phosphorylates and down-regulates FXR1, thereby regulating behavior dimensions related to mood disorders in humans.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST74180

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jun Qian et al.
Proceedings of the National Academy of Sciences of the United States of America, 112(11), 3469-3474 (2015-03-04)
Aberrant expression of RNA-binding proteins has profound implications for cellular physiology and the pathogenesis of human diseases such as cancer. We previously identified the Fragile X-Related 1 gene (FXR1) as one amplified candidate driver gene at 3q26-29 in lung squamous
Hannah M Phelps et al.
Journal of pediatric surgery, 54(6), 1198-1205 (2019-03-22)
Wilms tumor (WT) is the most common childhood kidney cancer globally. Our prior unbiased proteomic screen of WT disparities revealed increased expression of Fragile X-Related 1 (FXR1) in Kenyan specimens where survival is dismal. FXR1 is an RNA-binding protein that
Keita Nakanishi et al.
Journal of thoracic disease, 14(9), 3245-3254 (2022-10-18)
Sarcopenia and its marker, the psoas muscle index (PMI), have attracted attention as prognostic factors for various types of cancers. The fragile X-related 1 (FXR1) gene is highly expressed in myocytes, and FXR1 overexpression is a candidate biomarker for poor
Thomas Del'Guidice et al.
Proceedings of the National Academy of Sciences of the United States of America, 112(33), E4610-E4619 (2015-08-05)
Inhibition of glycogen synthase kinase 3β (GSK3β) is a shared action believed to be involved in the regulation of behavior by psychoactive drugs such as antipsychotics and mood stabilizers. However, little is known about the identity of the substrates through
Regina Fischer-Kešo et al.
Molecular and cellular biology, 34(23), 4244-4256 (2014-09-17)
Plakophilins 1 and 3 (PKP1/3) are members of the arm repeat family of catenin proteins and serve as structural components of desmosomes, which are important for cell-cell-adhesion. In addition, PKP1/3 occur as soluble proteins outside desmosomes, yet their role in

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service