Skip to Content
Merck
All Photos(6)

Key Documents

HPA014597

Sigma-Aldrich

Anti-MARCH8 antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-Cellular modulator of immune recognition, Anti-E3 ubiquitin-protein ligase MARCH8, Anti-MARCH-VIII, Anti-Membrane-associated RING finger protein 8, Anti-Membrane-associated RING-CH protein VIII, Anti-RING finger protein 178, Anti-c-MIR

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
€544.00

€544.00


Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)


Select a Size

Change View
100 μL
€544.00

About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

€544.00


Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

technique(s)

immunohistochemistry: 1:50-1:200
western blot: 0.04-0.4 μg/mL

immunogen sequence

VQCKVYVQLWKRLKAYNRVIYVQNCPETSKKNIFEKSPLTEPNFENKHGHGICHSDTNSSCCTEPEDTGAEIIHV

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... MARCH8(220972)

General description

Membrane-associated RING-CH 8 (MARCH8) is a member of the MARCH family of transmembrane proteins, which function as ubiquitin ligases. It has the RING-CH domain in its N-terminal which faces the cytosol. Its C-terminal contains a six amino acid domain, which is conserved across many species. This protein has a wide range of tissue expression. It is localized to the cell membrane and endosomes.

Immunogen

E3 ubiquitin-protein ligase MARCH8 recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

Membrane-associated RING-CH 8 (MARCH8) is involved in endosomal trafficking and the regulation of the expression of their substrates at the cell surface. It ubiquitinates tumor necrosis factor-related apoptosis inducing ligand (TRAIL) receptor 1, and decreases its expression and thus, controls its steady-state expression. It recognizes and binds with the transmembrane domain of transferrin receptor (TfR), and ubiquitinates and down-regulates it. MARCH8 also determines the fate of the clathrin-independent cargo, as well as their recycling and degradation. When expressed, it ubiquitinates and directs CD98 to lysosomes and late endosomes. It ubiquitinates and degrades IL-1 receptor accessory protein (IL1RAP), which leads to the down-regulation of IL-1β and the related pathways such as, NF-κB. It also regulates the turnover of CD81 and CD44, which are ubiquitously expressed proteins.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST72948

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Rui Chen et al.
Proceedings of the National Academy of Sciences of the United States of America, 109(35), 14128-14133 (2012-08-21)
The proinflammatory cytokine interleukin-1 (IL-1) signals via type I IL-1 receptor (IL-1RI) and IL-1 receptor accessory protein (IL1RAP), which leads to activation of the transcription factor NF-κB and induction of a range of downstream proteins involved in inflammatory and immune
Hideaki Fujita et al.
Journal of cell science, 126(Pt 13), 2798-2809 (2013-04-23)
The transferrin receptor (TfR) mediates the uptake of transferrin (Tf)-bound iron from the plasma into the cells of peripheral tissues. The TfR continuously recycles between the plasma membrane and early/recycling endosomes. TfR expression is tightly controlled by the intracellular iron
Eric Bartee et al.
PloS one, 5(12), e15132-e15132 (2010-12-15)
Membrane-associated RING-CH (MARCH) proteins represent a family of transmembrane ubiquitin ligases modulating intracellular trafficking and turnover of transmembrane protein targets. While homologous proteins encoded by gamma-2 herpesviruses and leporipoxviruses have been studied extensively, limited information is available regarding the physiological
Craig A Eyster et al.
Molecular biology of the cell, 22(17), 3218-3230 (2011-07-16)
Following endocytosis, internalized plasma membrane proteins can be recycled back to the cell surface or trafficked to late endosomes/lysosomes for degradation. Here we report on the trafficking of multiple proteins that enter cells by clathrin-independent endocytosis (CIE) and determine that
Bert van de Kooij et al.
The Journal of biological chemistry, 288(9), 6617-6628 (2013-01-10)
The eleven members of the membrane-associated RING-CH (MARCH) ubiquitin ligase family are relatively unexplored. Upon exogenous (over)expression, a number of these ligases can affect the trafficking of membrane molecules. However, only for MARCH-1 endogenous functions have been demonstrated. For the

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service