Skip to Content
Merck
All Photos(7)

Documents

ZMS1121

Sigma-Aldrich

Anti-CMV immediate early Antibody, clone 6F8.2 ZooMAb® Mouse Monoclonal

newenhanced validation
greener alternative

recombinant, expressed in HEK 293 cells

Synonym(s):

HCMV immediate-early (IE) proteins, IE1, IE2, Optimized for rapid cell lysis and RNA stabilization for direct analysis, UL122

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203

biological source

mouse

Quality Level

recombinant

expressed in HEK 293 cells

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

6F8.2, recombinant monoclonal

description

recombinant, expressed in HEK 293 cells

product line

ZooMAb® learn more

form

lyophilized

mol wt

calculated mol wt 60-80 kDa
observed mol wt ~60-80 kDa

purified by

using protein G

species reactivity

virus

packaging

antibody small pack of 25

greener alternative product characteristics

Waste Prevention
Designing Safer Chemicals
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

sustainability

Greener Alternative Product

technique(s)

immunocytochemistry: suitable
immunohistochemistry: suitable
western blot: suitable

isotype

IgG2a

epitope sequence

Unknown

UniProt accession no.

greener alternative category

shipped in

ambient

storage temp.

2-8°C

General description

We are committed to bringing you greener alternative products, which adhere to one or more of the 12 Principles of Green Chemistry. This antibody is preservative-free, produced without the harm or sacrifice of animals and exceptionally stable to allow for ambient shipping and storage if needed, and thus aligns with "Waste Prevention", "Designing Safer Chemicals" and "Design for Energy Efficiency". Click here for more information.
ZooMAb® antibodies represent an entirely new generation of recombinant monoclonal antibodies. Each ZooMAb® antibody is manufactured using our proprietary recombinant expression system, purified to homogeneity, and precisely dispensed to produce robust and highly reproducible lot-to-lot consistency. Only top-performing clones are released for use by researchers. Each antibody is validated for high specificity and affinity across multiple applications, including its most commonly used application. ZooMAb® antibodies are reliably available and ready to ship when you need them.

Specificity

Clone 6F8.2 is a ZooMAb® mouse recombinant monoclonal antibody that detects CMV immediate early proteins.

Immunogen

Affinity purified CMV early antigen from MRC-5 cells infected with CMV AD169.

Application

Quality Control Testing

Evaluated by Immunohistochemistry (Paraffin) in MRC-5 cells infected with CMV..


Immunohistochemistry (Paraffin) Analysis: A 1:1,000 Dilution of this antibody detected CMV immediate early in MRC-5 cells infected with CMV.

Tested Applications

Western Blotting Analysis: A 1:1,000 dilution from a representative lot detected CMV immediate early in IMR-32 and SH-SY5Y cell lysates.

Immunocytochemistry Analysis: A 1:100 dilution from a representative lot detected CMV immediate early in MRC-5 cells infected with CMV.

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user.

Target description

Cytomegalovirus immediate early proteins (IE) are produced by the human cytomegalovirus (HCMV). HCMV belongs to the Orthoherpesviridae family and is known to infect a wide range of cell types. IE proteins expressed during the immediate early phase of viral infection. IE proteins are critical in the early stages of viral infection and reactivation from viral latency. IE antigen (alpha gene expression) are expressed at 3-12 hrs post-infection. Other CMV antigens are expressed at different times. Early Antigen (beta genes) a.k.a. Delayed Early or Intermediate Early: expressed at 12-24 hrs post-infection. Late Antigen (gamma genes) are expressed at 36-48hrs post-infection. IE can be detected in the nucleus of infected cells, where it interacts with host cell proteins and viral DNA. The expression of the IE protein is tightly regulated and essential for viral replication. HCMV infection is associated with various clinical manifestations, including congenital infections, complications in immunocompromised individuals, and organ transplant rejection. This ZooMAb® recombinant monoclonal antibody, generated by our proprietary technology, offers significantly enhanced specificity, affinity, reproducibility, and stability over conventional monoclonals.

Physical form

Purified recombinant mouse monoclonal antibody IgG, lyophilized in PBS with 5% Trehalose, normal appearance a coarse or translucent resin. The PBS/trehalose components in the ZooMAb formulation can have the appearance of a semi-solid (bead like gel) after lyophilization. This is a normal phenomenon. Please follow the recommended reconstitution procedure in the data sheet to dissolve the semi-solid, bead-like, gel-appearing material. The resulting antibody solution is completely stable and functional as proven by full functional testing. Contains no biocide or preservatives, such as azide, or any animal by-products. Larger pack sizes provided as multiples of 25 µL.

Reconstitution

300 µg/mL after reconstitution at 25 µL per vial. Please refer to guidance on suggested starting dilutions and/or titers per application and sample type.

Storage and Stability

Recommend storage of lyophilized product at 2-8°C; Before reconstitution, micro-centrifuge vials briefly to spin down material to bottom of the vial; Reconstitute each vial by adding 25 µL of filtered lab grade water or PBS; Reconstituted antibodies can be stored at 2-8°C, or -20°C for long term storage. Avoid repeated freeze-thaws.

Legal Information

ZooMAb is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service