Skip to Content
Merck
All Photos(5)

Key Documents

HPA009985

Sigma-Aldrich

Anti-PIK3CA antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-PI3-kinase p110 subunit α antibody produced in rabbit, Anti-PI3K antibody produced in rabbit, Anti-Phosphatidylinositol-4,5-bisphosphate 3-kinase catalytic subunit α-isoform antibody produced in rabbit, Anti-PtdIns-3-kinase p110 antibody produced in rabbit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunofluorescence: 1-4 μg/mL
immunohistochemistry: 1:50- 1:200

immunogen sequence

EDLLNPIGVTGSNPNKETPCLELEFDWFSSVVKFPDMSVIEEHANWSVSREAGFSYSHAGLSNRLARDNELRENDKEQLKAISTRDPLSEITEQEKDFLWSHRHYCVTIPEILPKLLLSVK

UniProt accession no.

application(s)

research pathology

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... PIK3CA(5290)

General description

PIK3CA (phosphatidylinositol-4,5-bisphosphate 3-kinase catalytic subunit α) is one of the four subunits of the 110kDa catalytic domain of PI3K protein, which contains a regulatory region of 85kDa. PI3K proteins form a family of lipid kinases. This gene is localized to human chromosome 3q.

Immunogen

Phosphatidylinositol-4,5-bisphosphate 3-kinase catalytic subunit α-isoform recombinant protein epitope signature tag (PrEST)

Application

Anti-PIK3CA antibody produced in rabbit, a Prestige Antibody, is developed and validated by the Human Protein Atlas (HPA) project . Each antibody is tested by immunohistochemistry against hundreds of normal and disease tissues. These images can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. The antibodies are also tested using immunofluorescence and western blotting. To view these protocols and other useful information about Prestige Antibodies and the HPA, visit sigma.com/prestige.

Biochem/physiol Actions

PIK3CA (phosphatidylinositol-4,5-bisphosphate 3-kinase catalytic subunit α) is one of the four catalytic subunits of PI3K protein, which plays an essential role in extracellular growth signaling. It phosphorylates and activates protein kinase B (PKB) protein. This protein has a wide range of physiological functions, and plays central role in multiple types of cancers. PIK3CA gene is either mutated or amplified in brain, breast, stomach, liver, ovary and colon cancers. Mutation in this gene leads to enhanced PI3K activity, and it is a candidate transforming oncogene. It is over-expressed in invasive breast cancer and is a marker for poor prognosis. This gene is amplified in high-grade dysplastic lesions from bronchial biopsy specimens obtained from preinvasive squamous lung cancer. Frequency of mutations in PIK3CA gene is higher in tumors without lymph node metastasis, than those within gastric cancer. Thus, it might be essential to evaluate the mutations in this gene to determine cetuximab treatment in gastric cancer patients.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST86825.

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mohammed A Aleskandarany et al.
Breast cancer research and treatment, 122(1), 45-53 (2009-08-25)
The implications of Phosphatidylinositol 3-kinase (PIK3CA) mutations and its aberrant protein expression in breast cancer (BC) different molecular subtypes and patients' outcome remain controversial. The aims of this study were to assess the prevalence and clinical significance of PIK3CA protein
Frank McCaughan et al.
American journal of respiratory and critical care medicine, 182(1), 83-91 (2010-03-20)
Amplification of distal 3q is the most common genomic aberration in squamous lung cancer (SQC). SQC develops in a multistage progression from normal bronchial epithelium through dysplasia to invasive disease. Identifying the key driver events in the early pathogenesis of
Piotr Laudański et al.
Folia histochemica et cytobiologica, 49(2), 317-324 (2011-07-12)
The aim of this study was to selectively profile the activation status of mammalian target of rapamycin (mTOR)-associated oncogenes and tumor suppressor genes (TSGs) in ovarian cancer specimens, healthy ovaries and benign ovarian tumors, including endometrial cysts. We used a
Maider Ibarrola-Villava et al.
Oncotarget, 6(29), 26935-26945 (2015-09-04)
Genetic and epigenetic alterations play an important role in gastric cancer (GC) pathogenesis. Aberrations of the phosphatidylinositol-3-kinase signaling pathway are well described. However, emerging genes have been described such as, the chromatin remodeling gene ARID1A. Our aim was to determine
Si-Hyong Jang et al.
Journal of gastric cancer, 16(2), 85-92 (2016-07-20)
PIK3CA is often mutated in a variety of malignancies, including colon, gastric, ovary, breast, and brain tumors. We investigated PIK3CA expression in gastric cancer and explored the relationships between the PIK3CA expression level and clinicopathological features as well as survival

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service