Skip to Content
Merck
All Photos(6)

Key Documents

HPA009178

Sigma-Aldrich

Anti-CD79B antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-B-cell antigen receptor complex-associated protein β-chain precursor antibody produced in rabbit, Anti-B-cell-specific glycoprotein B29 antibody produced in rabbit, Anti-CD79b antigen antibody produced in rabbit, Anti-IG-β antibody produced in rabbit, Anti-Immunoglobulin-associated B29 protein antibody produced in rabbit

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
€505.00

€505.00


Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)


Select a Size

Change View
100 μL
€505.00

About This Item

MDL number:
UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

€505.00


Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

orthogonal RNAseq
recombinant expression
Learn more about Antibody Enhanced Validation

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:200-1:500

immunogen sequence

DKDDSKAGMEEDHTYEGLDIDQTATYEDIVTLRTGEVKWSVGEHPGQE

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... CD79B(974)

General description

CD79B (cluster of differentiation 79β), along with CD79α, forms the transmembrane heterodimeric signaling part of the BCR (B-cell receptor) complex. This gene is localized to human chromosome 17.

Immunogen

B-cell antigen receptor complex-associated protein β-chain precursor recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

CD79B (cluster of differentiation 79β) forms an essential component of BC (B-cell receptor) signaling pathway. It is important for the initiation of the signaling cascade upon interaction of the BCR with antigen. Its expression in absent in terminally differentiated plasma cells and its aberrant expression is linked with various B-cell malignancies, such as chronic lymphocytic leukemia. Mutations in this gene are involved in the pathogenesis of chronic lymphocytic leukemia (CLL). CD79B plays an essential role in the development of human B cells and mutations in this gene can result in agammaglobulinemia.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST71612

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Isabela Cajiao et al.
American journal of hematology, 82(8), 712-720 (2007-02-23)
The etiology of chronic lymphocytic leukemia (CLL) is poorly understood and its course is highly variable. Somatic hypermutation (SHM) of the immunoglobulin heavy chain (IgV(H)) gene and ZAP70 protein expression have been reported as prognostic indicators. However, these assays are
Le Cai et al.
Oncology letters, 13(1), 51-56 (2017-01-27)
The v-Crk avian sarcoma virus CT10 oncogene homolog-like (CRKL) protein is important in cancer progression. However, its expression pattern and biological roles in human endometrial carcinoma remain unexplored. The potential mechanism of CRKL-induced cancer progression is still unclear. The present
Eung Jae Yoo et al.
The Journal of biological chemistry, 288(25), 18353-18365 (2013-05-08)
The human B cell-specific protein, CD79b (also known as Igβ and B29) constitutes an essential signal transduction component of the B cell receptor. Although its function is central to the triggering of B cell terminal differentiation in response to antigen
Simona Ferrari et al.
The Journal of experimental medicine, 204(9), 2047-2051 (2007-08-22)
Agammaglobulinemia is a rare primary immunodeficiency characterized by an early block of B cell development in the bone marrow, resulting in the absence of peripheral B cells and low/absent immunoglobulin serum levels. So far, mutations in Btk, mu heavy chain

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service