Skip to Content
Merck

Skip To

E5905

pCMV-BICEP-4 Expression Vector

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
20 μg
Check Cart for Availability
€ 811,00

About This Item

NACRES:
NA.56
UNSPSC Code:
41106621

€ 811,00


Check Cart for Availability

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


shipped in

dry ice

Quality Segment

storage temp.

−20°C

Application

For transient, cytoplasmic expression of N-terminal FLAG® fusions and N-terminal c-myc fusions from bicistronic mRNA. Two genes can be cloned into MCS1 and MCS2 for transcription of a single message from the CMV promoter. This vector is useful for protein-protein interaction studies and multi-subunit proteins. Supplied with a control vector (pCMV-BICEP-4+p53/LTA) containing FLAG-p53 and c-myc-Large T antigen in MCS1 and MCS2.

Other Notes

Browse additional application references in our FLAG® Literature portal.

Legal Information

FLAG is a registered trademark of Merck KGaA, Darmstadt, Germany
pCMV-BICEP is a trademark of Sigma-Aldrich Co. LLC

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
P9743P1243E7029
shipped in

dry ice

shipped in

dry ice

shipped in

dry ice

shipped in

dry ice

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C

Quality Level

200

Quality Level

200

Quality Level

200

Quality Level

200


Storage Class

10 - Combustible liquids



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters