Skip to Content
Merck
All Photos(2)

Key Documents

HPA008622

Sigma-Aldrich

Anti-CSMD2 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-CUB and sushi domain-containing protein 2 antibody produced in rabbit, Anti-CUB and sushi multiple domains protein 2 antibody produced in rabbit

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
€544.00

€544.00


Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)


Select a Size

Change View
100 μL
€544.00

About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

€544.00


Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunohistochemistry: 1:200- 1:500

immunogen sequence

RYSAPYCSLPRAPLHGFILGQTSTQPGGSIHFGCNAGYRLVGHSMAICTRHPQGYHLWSEAIPLCQALSCGLPEAPKNGMVFGKEYTVGTKAMYSCSEGYHLQAGAEATAECLDTGLWSNRNVPPQCVPVTCPDVSSISVEH

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... CSMD2(114784)

General description

CSMD2 (CUB and Sushi multiple domains 2) belongs to the three member family called CSMD. It is characterized by 14 CUB domains interspersed with sushi domain. These are followed by multiple consecutive sushi domains, one transmembrane domain, and a short cytosolic tail. This protein has a molecular weight of 383kDa. This gene is localized to human chromosome 1p35.1-34.3.

Immunogen

CUB and sushi domain-containing protein 2 recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

CSMD2 (CUB and Sushi multiple domains 2) is down-regulated in colorectal cancer (CRC), which is linked with tumor size and lymph invasion, and differentiation. Inactivation of this gene in CRC patients is linked with poor prognosis. Therefore, it is thought to be a tumor suppressor gene. Polymorphisms in this gene are linked with the etiology of schizophrenia. Its expression is increased in certain head and neck cancer cell lines, though it is localized to the chromosomal locus containing a suppressor of oligodendrogliomas.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST71865

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Rui Zhang et al.
Tumour biology : the journal of the International Society for Oncodevelopmental Biology and Medicine, 35(5), 4419-4423 (2014-01-11)
CUB and sushi multiple domain protein 1 (CSMD1) is a candidate tumor suppressor gene. The three members of CSMD family have very similar structures, each consisting of 14 CUB domains separated from one another by a sushi domain, an additional
Wei Ling Lau et al.
Genomics, 82(3), 412-415 (2003-08-09)
CSMD1 is a putative suppressor of squamous cell carcinomas mapping to human chromosomal region 8p23. We have cloned two new members of this gene family, CSMD2 and CSMD3. The three CSMD proteins have very similar structures, each consisting of 14
Bjarte Håvik et al.
Biological psychiatry, 70(1), 35-42 (2011-03-29)
Patients with schizophrenia often suffer from cognitive dysfunction, including impaired learning and memory. We recently demonstrated that long-term potentiation in rat hippocampus, a mechanistic model of learning and memory, is linked to gene expression changes in immunity-related processes involved in

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service