Skip to Content
Merck
All Photos(7)

Key Documents

HPA005809

Sigma-Aldrich

Anti-PRDM2 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-GATA-3-binding protein G3B antibody produced in rabbit, Anti-MTB-ZF antibody produced in rabbit, Anti-MTE-binding protein antibody produced in rabbit, Anti-PR domain zinc finger protein 2 antibody produced in rabbit, Anti-PR domain-containing protein 2 antibody produced in rabbit, Anti-Retinoblastoma protein-interacting zinc-finger protein antibody produced in rabbit, Anti-Zinc finger protein RIZ antibody produced in rabbit

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
€544.00

€544.00


Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)


Select a Size

Change View
100 μL
€544.00

About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

€544.00


Usually ships in 1 week. (Orders outside of US and Europe, please allow an additional 1-2 weeks for delivery)

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:50-1:200

immunogen sequence

PPPFQYHHRNPMGIGVTATNFTTHNIPQTFTTAIRCTKCGKGVDNMPELHKHILACASASDKKRYTPKKNPVPLKQTVQPKNGVVVLDNSGKNAFRRMGQPKRLNFSVELSKMSSNKLKLNALKKKNQLVQKAI

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... PRDM2(7799)

Immunogen

PR domain zinc finger protein 2 recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

PR domain containing 2, with ZNF domain (PRDM2) is referred as RIZ, KMT8, RIZ1, RIZ2, MTB-ZF and HUMHOXY1. The protein is encoded by the PRDM2 gene in humans. It is a 250-kDa nuclear protein containing eight zinc-finger motifs. This gene contains an Rb-binding motif sharing an antigenic epitope with the C terminus of E1A. It may have a role in transcriptional regulation during neuronal differentiation and pathogenesis of retinoblastoma. It is a tumor suppressor gene that is highly inactivated by promoter hypermethylation in patients with liver fluke-related cholangiocarcinoma (CCA). It might play a role in regulating cell proliferation and migration in CCA. Inactivation of this gene is related to the development and progression of esophageal cancer. Its expression is significantly down-regulated as the formation of meningiomas progresses and may be a promising candidate tumor suppressor gene that contributes to malignant meningiomas.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST70896

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Shang-Wen Dong et al.
World journal of gastroenterology, 19(37), 6170-6177 (2013-10-12)
To investigate the effect of retinoblastoma protein-interacting zinc finger gene 1 (RIZ1) upregulation in gene expression profile and oncogenicity of human esophageal squamous cell carcinoma (ESCC) cell line TE13. TE13 cells were transfected with pcDNA3.1(+)/RIZ1 and pcDNA3.1(+). Changes in gene
Z Y Liu et al.
Oncogene, 32(10), 1216-1222 (2012-05-23)
Retinoblastoma protein-interacting zinc-finger gene 1 (RIZ1) expression is often silenced in many types of human tumors. However, the relationship between RIZ1 expression and malignant meningiomas remains unclear. Here we have found for the first time that the expression of RIZ1
I M Buyse et al.
Proceedings of the National Academy of Sciences of the United States of America, 92(10), 4467-4471 (1995-05-09)
The retinoblastoma protein (Rb) is a target of viral oncoproteins. To explore the hypothesis that viral proteins may be structural mimics of cellular proteins, we have searched cDNA libraries for Rb-binding proteins. We report here the cloning of a cDNA
Prasong Khaenam et al.
Asian Pacific journal of cancer prevention : APJCP, 13(8), 4007-4011 (2012-10-27)
Retinoblastoma-interacting zinc finger gene (RIZ1) is a tumor suppressor gene which is highly inactivated by promoter hypermethylation in patients with liver fluke-related cholangiocarcinoma (CCA). Epigenetic aberration of this gene might withdraw the ability to restrain tumor cell proliferation and migration.
Shang-Wen Dong et al.
World journal of gastroenterology, 18(6), 576-582 (2012-03-01)
To investigate the promoter region methylation status of retinoblastoma protein-interacting zinc finger gene 1 (RIZ1) in the human esophageal squamous cell carcinoma (ESCC) cell lines and tissues and verify the relationship between methylation of RIZ1 and oncogenesis, tumor progression and

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service