Skip to Content
Merck
All Photos(4)

Key Documents

910937

Sigma-Aldrich

di-phenylethylammonium lead(II) tetraiodide

greener alternative

n = 1

Synonym(s):

(PEA)2PbI4, 2D perovskite, bis[(2-phenylethyl)ammonium] tetraiodoplumbate

Sign Into View Organizational & Contract Pricing

Select a Size

2.5 G
€538.00

€538.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
2.5 G
€538.00

About This Item

Empirical Formula (Hill Notation):
C16H24N2PbI4
CAS Number:
Molecular Weight:
959.19
MDL number:
UNSPSC Code:
12352101
NACRES:
NA.23

€538.00


Check Cart for Availability

Request a Bulk Order

form

crystals

greener alternative product characteristics

Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

greener alternative category

SMILES string

[N+H3]CCc2ccccc2.[N+H3]CCc1ccccc1

InChI key

PSWSOAQTFFDDBM-UHFFFAOYSA-P

General description

We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product has been enhanced for "Energy efficiency".

Application

Organic-inorganic hybrid perovskites based semiconducting materials exhibit excellent properties such as long charge carrier lifetime and diffusion length, and high photolumiscence quantum yield. These properties render the perovskite materials useful for applications in light emitting diodes (LEDs), lasers and photodetectors. The Bis[(2-phenylethyl)ammonium] tetraiomoplumbate is 2D-layered perovskite and finds applications in semiconductors, copper (II) probing, and in LEDs.[1][2][3][4]

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Repr. 1A - STOT RE 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ruddlesden?Popper Hybrid Lead Iodide Perovskite 2D Homologous Semiconductors.
Stoumpos C C, et al
Chemistry of Materials, 28, 2852-2867 (2016)
Chunqing Ma et al.
Chemical communications (Cambridge, England), 54(45), 5784-5787 (2018-05-22)
Stabilization of two-dimensional (2D) PEA2PbI4 (PEA is phenethylammonium) perovskite nanocrystals (PNCs) in water is achieved. By inhibiting the desorption process, the PNCs show exceptional stability for more than 2 months in PEA+ aqueous solutions. Stabilized PNCs are successfully applied for
Giant Two-Photon Absorption and Its Saturation in 2D Organic?Inorganic Perovskite.
Liu W, et al.
Advanced Optical Materials, 5, 1601045-1601045 (2017)
Shuang Yang et al.
Angewandte Chemie (International ed. in English), 56(15), 4252-4255 (2017-03-16)
Two-dimensional (2D) organic-inorganic hybrid perovskite nanosheets (NSs) are attracting increasing research interest due to their unique properties and promising applications. Here, for the first time, we report the facile synthesis of single- and few-layer free-standing phenylethylammonium lead halide perovskite NSs

Articles

Dr. Perini and Professor Correa-Baena discuss the latest research and effort to obtain higher performance and stability of perovskite materials.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service