Skip to Content
Merck
All Photos(2)

Key Documents

MAB2600

Sigma-Aldrich

Anti-Glypican-1 Antibody, clone 4D1

clone 4D1, from mouse

Synonym(s):

glypican 1, glypican proteoglycan 1

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€515.00

€515.00


Estimated to ship on25 May 2025


Request a Bulk Order

Select a Size

Change View
100 μG
€515.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€515.00


Estimated to ship on25 May 2025


Request a Bulk Order

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

4D1, monoclonal

species reactivity

human, rat

technique(s)

immunohistochemistry: suitable (paraffin)
western blot: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... GPC1(2817)
rat ... Gpc1(58920)

General description

Heparan sulfate proteoglycans (HSPGs) can be divided into general categories of
cell-associated forms, including syndecans, glypicans, and secreted extracellular matrix forms, such as perlecan. Glypicans contain six family members to date and are proteins bound to the outer surface of the plasma membrane by a glycosyl-phosphatidylinositol anchor and involved in cellular adhesion and migration and function as accessory molecules for signaling receptors, (Filmus, 2008 & Gui, 2006). Current publications indicate the main function of membrane-attached glypicans is to regulate the signaling of Wnts, Hedgehogs, fibroblast growth factors and bone morphogenetic proteins (BMPs), (Filmus, 2008).
Glypican-1 (GPC1) is over-expressed in human gliomas and appears to have a greater capability of promoting FGF-2 signaling in some gliomas versus HSPGs in normal astrocytes. Additional observations suggest that GPC1 can impact the growth of pancreatic cancer cells by attenuating tumor angiogenesis, (Whipple, 2008).

Specificity

MAB2600 recognizes glypican-1.

Immunogen

Epitope: Full Length
Full length recombinant protein.

Application

Anti-Glypican-1 Antibody, clone 4D1 detects level of Glypican-1 & has been published & validated for use in IH(P) & WB.
Immunohistochemistry:
A 1:50 dilution of a previous lot detected strong cytoplasmic staining of tumor cells in formalin-fixed, paraffin-embedded gangliobastoma tissue sample. There was negative staining of normal, uninvolved cells.
Research Category
Cell Structure
Research Sub Category
ECM Proteins

Quality

Routinely evaluated by Western blot on human PC-12 cell lysate using 0.5 µg/mL of Anti-Glypican-1 at 1 mg/mL.

Target description

Recognizes Glypican-1 at ~ 59 kDa

Physical form

Format: Purified
Protein G Purified
Purified in 0.1 M Tris-Glycine (pH 7.4) 150 mM NaCl with 0.05% NaN3.

Storage and Stability

Stable for 1 year at 2-8ºC from date of receipt.

Analysis Note

Control
PC-12 cell lysate.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tongrong He et al.
Journal of cerebral blood flow and metabolism : official journal of the International Society of Cerebral Blood Flow and Metabolism, 42(10), 1920-1932 (2022-06-09)
Cerebrovascular effects of β-site amyloid precursor protein-cleaving enzyme 1 (BACE1) inactivation have not been systematically studied. In the present study we employed cultured human brain microvascular endothelial cells (BMECs), BACE1-knockout (BACE1-/-) mice and conditional (tamoxifen-induced) endothelium-specific BACE1-knockout (eBACE1-/-) mice to
Steffen Rickelt et al.
Matrix biology : journal of the International Society for Matrix Biology, 71-72, 10-27 (2018-05-08)
The diversity of extracellular matrix (ECM) proteins encoded in mammalian genomes and detected by proteomic analyses generates a need for well validated antibodies against these proteins. We present characterization of a large number of antibodies against ECM proteins, from both

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service