Skip to Content
Merck
All Photos(2)

Key Documents

H0542

Sigma-Aldrich

HISTOSETTE® I Tissue Processing/Embedding Cassettes

white

Sign Into View Organizational & Contract Pricing

Select a Size

1 CASE
€226.00

€226.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 CASE
€226.00

About This Item

UNSPSC Code:
41116124
NACRES:
NB.17

€226.00


Check Cart for Availability

Request a Bulk Order

color

white

application(s)

hematology
histology

storage temp.

room temp

Looking for similar products? Visit Product Comparison Guide

General description

Histology processing/embedding cassettes made of acetal polymer. These cassettes keep specimens safely submerged in liquid and are resistant to histological solvents. The one-piece lid snaps apart and conveniently locks into the cassette base, ensuring specimen safety. A line of cassette colors are available to fit your needs and preference. A large 30° anterior writing area is available for your convenience. Not suitable for automated printers.

Quantity

Case contains 3 dispenser boxes of 500 cassettes.

Legal Information

Histosette is a registered trademark of Simport Plastics Ltd.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service