Skip to Content
Merck
All Photos(1)

Key Documents

GW22692

Sigma-Aldrich

Anti-CD55 antibody produced in chicken

affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-CD55

Sign Into View Organizational & Contract Pricing

Select a Size

50 μG
€289.00

€289.00


Usually ships in 2 weeks.

Request a Bulk Order

Select a Size

Change View
50 μG
€289.00

About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

€289.00


Usually ships in 2 weeks.

Request a Bulk Order

biological source

chicken

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

human

manufacturer/tradename

Genway 15-288-22692

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... CD55(1604)

Immunogen

Immunogen Sequence: GI # 10835143, sequence 36-129
Recombinant decay accelerating factor for complement

Application

Anti-CD55 antibody produced in chicken is suitable for western blotting analysis at a dilution of 1:500, for tissue or cell staining at a dilution of 1:200.

Biochem/physiol Actions

Complement decay-accelerating factor (decay-accelerating factor, DAF) is a protein encoded by the CD55 gene in humans. It plays multiple functions including tissue protection from the cytotoxic complement injury and an anti-inflammatory function due to its anti-adherence properties. It is essential in many organ systems including pregnancy for protection of the semiallogeneic fetus or for preventing uncontrolled infiltration by white cells in their pro- and/or anti-inflammatory functions. Expression of CD55 appears to have multiple regulatory tissue-specific and/or menstrual cycle-specific mechanisms that are involved in complex signaling mechanisms. Its regulation may involve a direct or an indirect effect of at least the estrogen, progesterone, and corticosteroid regulatory pathways. It plays a a primary role in conferring resistance to lysis. CD55 is overexpressed in several types of cancer.

Physical form

Solution in phosphate buffered saline containing 0.02% sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yanxia Wu et al.
American journal of physiology. Gastrointestinal and liver physiology, 306(12), G1056-G1064 (2014-04-26)
Engineering cancer cells to express heterologous antigen α-gal and induce the destruction of tumor cells depending on the complement cascade may be a promising strategy of tumor therapy. However, the feasibility and effect of using α-gal to induce colorectal adenocarcinoma
Bogdan Nowicki et al.
Advances in experimental medicine and biology, 735, 83-96 (2013-02-14)
In this chapter, we present a concise historic prospective and a summary of accumulated knowledge on steroid hormones, DAF expression, and therapeutic implication of steroid hormone treatment on multiple pathologies, including infection and the host-pathogen interactions. DAF/CD55 plays multiple physiologic
E D Vainer et al.
Tissue antigens, 82(1), 26-34 (2013-05-23)
CD55 (decay-accelerating factor, DAF) is overexpressed in several types of cancer, including colorectal cancer. Because of its inhibitory effect on the complement system, it has been suggested as a possible target for cancer immunotherapy. However, CD55 is also expressed in

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service