Skip to Content
Merck
All Photos(1)

Key Documents

GW22681

Sigma-Aldrich

Anti-MUC1 antibody produced in chicken

affinity isolated antibody, buffered aqueous solution

Sign Into View Organizational & Contract Pricing

Select a Size

50 μG
€289.00

€289.00


Check Cart for Availability
A recombinant, preservative-free antibody is available for your target. Try ZRB1016

Request a Bulk Order

Select a Size

Change View
50 μG
€289.00

About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

€289.00


Check Cart for Availability
A recombinant, preservative-free antibody is available for your target. Try ZRB1016

Request a Bulk Order

biological source

chicken

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

human, mouse, rat

manufacturer/tradename

Genway 15-288-22681

technique(s)

immunohistochemistry: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... MUC1(4582)

General description

Mucin 1 (MUC1) is a O-glycosylated, membrane bound, transmembrane glycoprotein belonging to the mucin (MUC) gene family. It is expressed on the apical surface of mucosal epithelia in the lung, eye, breast and stomach. It consists of an N-terminal extracellular domain consisting of variable number tandem repeats (MUC1-TR) of 20 amino acids (aa) and a C-terminal domain which includes a 53aa extracellular region, a transmembrane domain (TM) and a 72aa cytoplasmic tail (MUC1CT).

Immunogen

Immunogen Sequence: GI # 65301117, sequence 199-244
Recombinant MUC1 mucin isoform 1

Application

Anti-MUC1 antibody produced in chicken is suitbale for immunohistochemistry and western blotting analysis at a dilution of 1:500, for tissue or cell staining at a dilution of 1:200.

Biochem/physiol Actions

Mucin 1, cell surface associated (MUC1), plays an important role in the cell signaling during cancer progression. It carries tumor-associated epitopes and is aberrantly expressed in tumors. MUC1 plays a crucial role in the development, progression of breast cancer and could promote the invasion ability of breast cancer cells. Mutation in the gene may result in progressive chronic kidney failure with a bland urinary sediment.

Physical form

Solution in phosphate buffered saline containing 0.02% sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Anthony J Bleyer et al.
Clinical journal of the American Society of Nephrology : CJASN, 9(3), 527-535 (2014-02-11)
The genetic cause of medullary cystic kidney disease type 1 was recently identified as a cytosine insertion in the variable number of tandem repeat region of MUC1 encoding mucoprotein-1 (MUC1), a protein that is present in skin, breast, and lung
S J Gendler et al.
The Journal of biological chemistry, 265(25), 15286-15293 (1990-09-05)
Human mammary cells present on the cell surface a polymorphic epithelial mucin (PEM) which is developmentally regulated and aberrantly expressed in tumors. PEM carries tumor-associated epitopes recognized by the monoclonal antibodies HMFG-1, HMFG-2, and SM-3. Previously isolated partial cDNA clones
M Sahraei et al.
Oncogene, 31(47), 4935-4945 (2012-01-24)
Pancreatic ductal adenocarcinoma (PDA) has one of the worst prognoses of all cancers. Mucin 1 (MUC1), a transmembrane mucin glycoprotein, is a key modulator of several signaling pathways that affect oncogenesis, motility and metastasis. Its expression is known to be
Lili Zhang et al.
The Journal of pathology, 234(1), 60-73 (2014-05-20)
Cigarette smoke increases the risk of lung cancer by 20-fold and accounts for 87% of lung cancer deaths. In the normal airway, heavily O-glycosylated mucin-1 (MUC1) and adherens junctions (AJs) establish a structural barrier that protects the airway from infectious
Ya-Wen Wang et al.
Journal of cancer research and clinical oncology, 140(8), 1271-1281 (2014-05-03)
Increasing evidences demonstrate that CD227 plays a crucial role in the development and progression of breast cancer. However, the function of CD227 in breast carcinoma was still controversial and the investigation on CD227 in Asian race was scarce. To investigate

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service