Skip to Content
Merck
All Photos(25)

Key Documents

ZRB1016

Sigma-Aldrich

Anti-MUC1 Antibody, clone 1F23 ZooMAb® Rabbit Monoclonal

enhanced validation
greener alternative

recombinant, expressed in HEK 293 cells

Synonym(s):

Breast carcinoma-associated antigen DF3, CA 15-3, CD227, Cancer antigen 15-3, Carcinoma-associated mucin, EMA, Episialin, H23AG, KL-6, Krebs von den Lungen-6, Mucin-1, PEM, PEMT, PUM, Peanut-reactive urinary mucin, Polymorphic epithelial mucin, Tumor-associated epithelial membrane antigen, Tumor-associated mucin

Sign Into View Organizational & Contract Pricing

Select a Size

25 μL
€163.00
4 X 25 μL
€336.00

€163.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 μL
€163.00
4 X 25 μL
€336.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

€163.00


Check Cart for Availability

Request a Bulk Order

biological source

rabbit

Quality Level

recombinant

expressed in HEK 293 cells

conjugate

unconjugated

antibody form

purified antibody

clone

1F23, recombinant monoclonal
recombinant monoclonal

product line

ZooMAb® learn more

form

lyophilized

mol wt

calculated mol wt 122.10 kDa

species reactivity

human, mouse

packaging

antibody small pack of 25 μL

greener alternative product characteristics

Waste Prevention
Designing Safer Chemicals
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

sustainability

Greener Alternative Product

technique(s)

affinity binding assay: suitable
flow cytometry: suitable
immunocytochemistry: suitable
immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable
western blot: suitable

isotype

IgG

UniProt accession no.

greener alternative category

shipped in

ambient

storage temp.

2-8°C

target post-translational modification

unmodified

General description

We are committed to bringing you greener alternative products, which adhere to one or more of The 12 Principles of Green Chemistry.This antibody is Preservative-free, produced without the harm or sacrifice of animals and exceptionally stable to allow for ambient shipping and storage if needed and thus aligns with "Waste Prevention", "Designing Safer Chemicals" and "Design for Energy Efficiency". Click here for more information.
ZooMAb antibodies represent an entirely new generation of recombinant monoclonal antibodies.

Each ZooMAb antibody is manufactured using our proprietary recombinant expression system, purified to homogeneity, and precisely dispensed to produce robust and highly reproducible lot-to-lot consistency. Only top-performing clones are released for use by researchers. Each antibody is validated for high specificity and affinity across multiple applications, including its most commonly used application. ZooMAb antibodies are reliably available and ready to ship when you need them.

Learn more about ZooMAb here.

Specificity

Clone 1F23 is a ZooMAb rabbit recombinant monoclonal antibody that specifically detects Mucin-1 (MUC1). It targets an epitope within 16 amino acids from the C-terminal, extracellular domain.

Immunogen

KLH-conjugated linear peptide corresponding to 16 amino acids from Human Mucin-1.

Application

Quality Control Testing
Immunohistochemistry (Paraffin) Analysis: A 1:1,000 dilution of this antibody detected Mucin-1 (MUC1) in human stomach tissue sections.Tested Applications Affinity Binding Assay: A representative lot of this antibody bound Mucin-1 (MUC1) peptide with a KD of 1.6 x 10-6 in an affinity binding assay.Immunocytochemistry Analysis: A 1:100 dilution from a representative lot detected Mucin-1 (MUC1) in HeLa, A431, HepG2, NIH 3T3 and MCF-7 cells.Western Blotting Analysis: A 1:1,000 dilution from a representative lot detected Mucin-1 (MUC1) in Mouse Pancreas tissue lysate.Flow Cytometry Analysis: 1 μg from a representative lot detected Mucin-1 (MUC1) in one million HeLa cells.Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user
Anti-MUC1, clone 1F23 ZooMAb, Cat. No. ZRB1016, is a recombinant Rabbit monoclonal antibody that targets Mucin-1 and is tested for use in Affinity Binding Assay, Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Paraffin), and Western Blotting.

Target description

Mucin-1 (UniProt: P15941; also known as MUC-1, Breast carcinoma-associated antigen DF3, Cancer antigen 15-3, CA 15-3, Carcinoma-associated mucin, Episialin, H23AG, Krebs von den Lungen-6, KL-6, PEMT, Peanut-reactive urinary mucin, PUM, Polymorphic epithelial mucin, PEM, Tumor-associated epithelial membrane antigen, EMA, Tumor-associated mucin, CD227) is encoded by the MUC1 (also known as PUM) gene (Gene ID: 4582) in human. Mucins are high molecular weight glycoproteins that consist of a mucin core protein and O-linked carbohydrates. Mucins are present on the surface of, and are secreted by, many glandular epithelial cells. They provide a barrier between the luminal membranes of cells in the epithelium and their environment. Mucins consist of a nonglobular protein backbone (apomucin) and many O-linked glycans. Mucin-1 is a highly glycosylated, single-pass type I membrane protein that is exclusively located in the apical domain of the plasma membrane of highly polarized epithelial cells and is internalized and recycled to cell membrane following endocytosis. Its expression has been reported in airway passages and in breast and uterine tissue. It is also expressed in activated and unactivated T-cells. It is overexpressed in epithelial tumors, such as breast and ovarian cancers. Mucin-1 is synthesized with a signal peptide (aa 1-23), which is subsequently removed to generate the mature form that contains an extracellular domain (aa 24-1158), a transmembrane domain (aa 1159-1181), and a cytoplasmic domain (aa 1182-1255). It consists of an alpha (aa 24-1097) and a beta (aa 1098-1255) subunit. The alpha subunit forms a tight, non-covalent heterodimeric complex with the proteolytically-released beta-subunit. The alpha subunit has cell adhesive properties. The beta subunit contains a C-terminal domain which is involved in cell signaling, through phosphorylations and protein-protein interactions. Phosphorylation on tyrosine in the C-terminal region increases its nuclear location. Phosphorylation by PKC delta induces binding of Mucin-1 to beta-catenin and thus decreases the formation of the beta-catenin/E-cadherin complex. Mutations in MUC1 have been linked to Medullary cystic kidney disease that is characterized by formation of renal cysts at the corticomedullary junction and impaired renal function. This ZooMAb recombinant monoclonal antibody, generated by our propriety technology, offers significantly enhanced specificity, affinity, reproducibility, and stability over conventional monoclonals.

Physical form

Purified recombinant rabbit monoclonal antibody IgG, lyophilized in PBS with 5% Trehalose, normal appearance a coarse or translucent resin. Contains no biocide or preservatives, such as azide, or any animal by-products. Larger pack sizes provided as multiples of 25 μL.

Reconstitution

Reconstitute lyophilized antibody pellet with 25μL of ultrapure water or Phosphate Buffered Saline (PBS). Please refer to our reconstitution protocol and the specific application guidance on the suggested starting dilutions and sample type.

Storage and Stability

Recommend storage of lyophilized product at 2-8°C; Before reconstitution, micro-centrifuge vials briefly to spin down material to bottom of the vial; Reconstitute each vial by adding 25 μL of filtered lab grade water or PBS; Reconstituted antibodies can be stored at 2-8°C, or -20°C for long term storage. Avoid repeated freeze-thaws.

Legal Information

ZooMAb is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service