Skip to Content
Merck
All Photos(1)

Key Documents

GW21222

Sigma-Aldrich

Anti-c-Myc Tag Antibody

chicken polyclonal

Synonym(s):

Anti-Myc proto-oncogene protein

Sign Into View Organizational & Contract Pricing

Select a Size

50 μG
€385.00

€385.00


Usually ships in 2 weeks.
A recombinant, preservative-free antibody is available for your target. Try ZRB1003

Request a Bulk Order

Select a Size

Change View
50 μG
€385.00

About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

€385.00


Usually ships in 2 weeks.
A recombinant, preservative-free antibody is available for your target. Try ZRB1003

Request a Bulk Order

Product Name

Anti-c-Myc (ab2) antibody produced in chicken, affinity isolated antibody, buffered aqueous solution

biological source

chicken

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

mouse, rat, human

manufacturer/tradename

Genway 15-288-21222

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... MYC(4609)

Immunogen

Immunogen Sequence: GI # 12962935, sequence 268-386
Recombinant V-myc myelocytomatosis viral oncogene homolog

Application

Anti-c-Myc (ab2) antibody produced in chicken is suitable for western blotting analysis at a dilution of 1:500, for tissue or cell staining at a dilution of 1:200.

Biochem/physiol Actions

Myc (c-Myc) is a regulatory gene and encodes for a transcription factor. It is a multifunctional, nuclear phosphoprotein that plays a role in cell cycle progression, apoptosis and cellular transformation. Myc is associated with aggressive behaviour in several cancers. c-MYC gain with lung adenocarcinoma is a candidate for additional first-line treatment to mitigate increased risk for disease progression and death. It plays an important role in the pathogenesis of B-cell prolymphocytic leukemia (B-PLL).

Physical form

Solution in phosphate buffered saline containing 0.02% sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ellen Flatley et al.
American journal of clinical pathology, 142(3), 347-354 (2014-08-16)
B-cell prolymphocytic leukemia (B-PLL) remains a controversial entity, and its molecular pathogenesis is largely unknown. Patients are older, typically having marked lymphocytosis and splenomegaly in the absence of lymphadenopathy. It is defined as a mature B-cell leukemia with more than
A N Seo et al.
British journal of cancer, 110(11), 2688-2699 (2014-05-09)
c-MYC copy number gain (c-MYC gain) has been associated with aggressive behaviour in several cancers. However, the role of c-MYC gain has not yet been determined in lung adenocarcinomas classified by genetic alterations in epidermal growth factor receptor (EGFR), KRAS

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service