Skip to Content
Merck
All Photos(1)

Documents

09859

Sigma-Aldrich

Ammonium hydroxide solution

BioUltra, ~1 M NH3 in H2O (T)

Synonym(s):

Ammonia aqueous, Ammonia water

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NH4OH
CAS Number:
Molecular Weight:
35.05
Beilstein:
3587154
MDL number:
UNSPSC Code:
12352305
eCl@ss:
38060103
PubChem Substance ID:
NACRES:
NA.25

product line

BioUltra

Quality Level

form

liquid

expl. lim.

27 %

concentration

~1 M NH3 in H2O (T)

impurities

insoluble matter, passes filter test

density

0.9 g/mL at 25 °C (lit.)

anion traces

chloride (Cl-): ≤0.5 mg/kg
phosphate (PO43-): ≤0.5 mg/kg
sulfate (SO42-): ≤2 mg/kg

cation traces

Al: ≤1 mg/kg
As: ≤0.1 mg/kg
Ba: ≤1 mg/kg
Bi: ≤1 mg/kg
Ca: ≤5 mg/kg
Cd: ≤1 mg/kg
Co: ≤1 mg/kg
Cr: ≤1 mg/kg
Cu: ≤1 mg/kg
Fe: ≤1 mg/kg
K: ≤20 mg/kg
Li: ≤1 mg/kg
Mg: ≤1 mg/kg
Mn: ≤1 mg/kg
Mo: ≤1 mg/kg
Na: ≤20 mg/kg
Ni: ≤1 mg/kg
Pb: ≤1 mg/kg
Sr: ≤1 mg/kg
Zn: ≤1 mg/kg

λ

neat

UV absorption

λ: 260 nm Amax: 0.01
λ: 280 nm Amax: 0.01

storage temp.

2-8°C

SMILES string

[NH4+].[OH-]

InChI

1S/H3N.H2O/h1H3;1H2

InChI key

VHUUQVKOLVNVRT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Ammonium hydroxide solution can be used to study chromatography, base solutions, analytical reagents, titration and volumetric solutions. Ammonium hydroxide solution has been used to develop a single-laboratory validation by inductively coupled plasma-mass spectrometry for the determination of total iodine (m/z 127) in infant formula and adult nutritional products. Ammonium hydroxide solution has also been used to develop a sensitive liquid chromatography-tandem mass spectrometry method to quantify miglustat in mouse plasma and in human plasma, which has improved treatment of patients with cystic fibrosis.

Pictograms

CorrosionEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 2 - Eye Dam. 1 - Skin Irrit. 2

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Y H Tak et al.
Analytical and bioanalytical chemistry, 401(10), 3275-3281 (2011-10-21)
A capillary electrophoresis method with UV-absorbance detection was studied and optimized for the determination of underivatized amino acids in urine. To improve concentration sensitivity the utility of in-capillary analyte stacking via dynamic pH junction was investigated with phenylalanine (Phe) and
Konstantin Chingin et al.
Analytical chemistry, 84(15), 6856-6862 (2012-07-12)
We introduce an online multiple-junction capillary isoelectric focusing fractionator (OMJ-CIEF) for separation of biological molecules in solution by pI. In OMJ-CIEF, the separation capillary is divided into seven equal sections joined with each other via tubular Nafion membrane insertions. Each
Ciceron Ayala-Orozco et al.
ACS nano, 8(6), 6372-6381 (2014-06-04)
Au nanoparticles with plasmon resonances in the near-infrared (NIR) region of the spectrum efficiently convert light into heat, a property useful for the photothermal ablation of cancerous tumors subsequent to nanoparticle uptake at the tumor site. A critical aspect of
Dominic J Hare et al.
ACS chemical neuroscience, 6(3), 398-402 (2015-01-15)
Plasma iron levels are decreased in Alzheimer's disease (AD) and associated with an idiopathic anemia. We examined iron-binding plasma proteins from AD patients and healthy controls from the Australian Imaging, Biomarkers and Lifestyle (AIBL) Flagship Study of Ageing using size
Marilia Freitas Calmon et al.
Colloids and surfaces. B, Biointerfaces, 100, 177-184 (2012-07-07)
Magnetic iron oxide nanoparticles (magnetite) (MNPs) were prepared using different organic and inorganic bases. Strong inorganic base (KOH) and organic bases (NH(4)OH and 1,4-diazabicyclo[2.2.2]octane (DABCO)) were used in the syntheses of the MNPs. The MNPs were characterized by X-ray diffraction

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service