GERPN201
Cell Proliferation Labelling Reagent
Cytiva RPN201
About This Item
Recommended Products
packaging
pkg of 10 mL
manufacturer/tradename
Cytiva RPN201
storage temp.
2-8°C
General description
Application
Features and Benefits
- Used in conjunction with RPN20, the reagent allows extended use of the kit where additional amounts of labeling reagent.
are required, for example, for in vivo studies.
Storage and Stability
Analysis Note
Storage Class Code
12 - Non Combustible Liquids
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service