Skip to Content
Merck
All Photos(1)

Key Documents

MABD55

Sigma-Aldrich

Anti-TERT (human) Antibody, clone 2C4

clone 2C4, from mouse

Synonym(s):

Telomerase reverse transcriptase, HEST2, Telomerase catalytic subunit, Telomerase-associated protein 2, Short name=TP2

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
€475.00

€475.00


Estimated to ship on30 May 2025



Select a Size

Change View
100 μL
€475.00

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

€475.00


Estimated to ship on30 May 2025


biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

2C4, monoclonal

species reactivity

human

technique(s)

immunocytochemistry: suitable
immunoprecipitation (IP): suitable
western blot: suitable

isotype

IgMκ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... TERT(7015)

General description

Human telomerase reverse transcriptase (hTERT) is the catalytic subunit of the telomerase. It is also responsible for addition of TTAGGG repeats to the telomere ends of the chromosome, which is important in maintaining cell immortality. In most normal human cells the expression of hTERT is very low. On the other hand, greater than 90 % of human tumors of different pathology and origins have been shown to upregulate telomerase expression, making hTERT a cancer-associated target.

Immunogen

Full length recombinant protein corresponding to human TERT.

Application

Anti-TERT (human) Antibody, clone 2C4 is an antibody against TERT (human) for use in WB, IC & IP.
Immunoprecipitation Analysis: A representative lot was used by an independent laboratory to detect TERT in HeLa cells (Masutomi, K., et al. (2003). Cell, Vol. 114:241–253).

Immunocytochemistry Analysis: A representative lot was used by an independent laboratory to detect TERT in HeLa cells, BJ and WI-38 fibroblasts (Masutomi, K., et al. (2003). Cell, Vol. 114:241–253).

Quality

Evaluated by Western Blot in human lymph node tissue lysate.

Western Blot Analysis: A 1:2,000 dilution of this antibody detected TERT in 10 µg of human lymph node tissue lysate.

Target description

~90 kDa observed.
Three isoforms exist for human TERT: Isoform 1 at 127 kDa, Isoform 2 at 90 kDa, and Isoform 3 at 120 kDa

Physical form

Format: Purified

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Andrew T Meram et al.
Anticancer research, 38(7), 3843-3852 (2018-07-05)
Hydrogen sulfide (H2S) and the enzymes that synthesize it, cystathionine-b-synthase, cystathionine γ-lyase, and 3-mercaptopyruvate, are increased in different human malignancies. Due to its short half-life, H2S concentrations have not been directly measured in a human malignancy. Here we directly measured

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service