Skip to Content
Merck
All Photos(1)

Key Documents

GE28950859

HiTrap® MacroCap SP

pkg of 5 × 5 mL, Cytiva, 28-9508-59

Sign Into View Organizational & Contract Pricing

Select a Size

5 X 5 ML
€2,260.00

€2,260.00


Estimated to ship on10 April 2025



Select a Size

Change View
5 X 5 ML
€2,260.00

About This Item

UNSPSC Code:
41106500
NACRES:
NB.21

€2,260.00


Estimated to ship on10 April 2025


packaging

pkg of 5 × 5 mL

manufacturer/tradename

28-9508-59
Cytiva

Legal Information

HiTrap is a registered trademark of Cytiva
MacroCap is a trademark of Cytiva

Signal Word

Danger

Hazard Statements

Hazard Classifications

Carc. 1B - Flam. Liq. 2 - Muta. 1B - Skin Sens. 1 - STOT RE 2 Oral

Target Organs

Peripheral nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH SVHC Candidate List

CAS No.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service