Skip to Content

PLA0083

Rabbit anti-FUS Antibody, Affinity Purified

Powered by Bethyl Laboratories, Inc.

Synonym(s):

75 kDa DNA-pairing protein, ALS6, ETM4, FUS1, HNRNPP2, POMP75, TLS, amyotrophic lateral sclerosis 6, derived from t(12;16) malignant liposarcoma, fus-like protein, fused in sarcoma, fusion, fusion (involved in t(12;16) in malignant liposarcoma), fusion gene in myxoid liposarcoma, heterogeneous nuclear ribonucleoprotein P2, hnRNP-P2, oncogene FUS, oncogene TLS, translocated in liposarcoma, translocated in liposarcoma protein

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NA.41
UNSPSC Code:
12352203
Conjugate:
unconjugated
Clone:
polyclonal
Application:
IHC, IP, WB
Citations:
1
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

rabbit

Quality Segment

conjugate

unconjugated

antibody form

affinity purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

species reactivity

mouse, human

technique(s)

immunohistochemistry: 1:1,000-1:5,000, immunoprecipitation (IP): 2-10 μg/mg, western blot: 1:2,000-1:10,000

accession no.

P35637

UniProt accession no.

shipped in

wet ice

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

rabbit ... FUS(2521)

Immunogen

The epitope recognized by PLA0083 maps to a region between residues 400 and 450 of human fusion (involved in t(12;16) in malignant liposarcoma) using the numbering given in SwissProt entry P35637 (GeneID 2521).

Physical form

Tris-citrate/phosphate buffer, pH 7 to 8 containing 0.09% Sodium Azide

Other Notes

FUS has been identified as a frequent translocation fusion partner of various transcription factors. FUS fusion genes have been shown to be associated with multiple tumor types which include liposarcoma, leukemia, histocytoma, and sarcoma. FUS is a multifunctional RNA-binding protein that associates with the nuclear matrix and Cajal bodies and appears to play a role in splicesome assembly, pre-mRNA splicing, DNA repair, transcriptional regulation and homologous recombination.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.


Still not finding the right product?

Try our Product Selector Tool to narrow your options


Storage Class

12 - Non Combustible Liquids

WGK

nwg

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

77-92-9

CAS No.


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library