GE28-9365-51
Hisprep™ Fast Flow 16/10
Cytiva 28-9365-51
Synonym(s):
16/10 Hisprep, Hisprep Column, Hisprep Fast Flow Column
About This Item
Recommended Products
packaging
pkg of 1 ea
manufacturer/tradename
Cytiva 28-9365-51
parameter
1.5 bar (22 psi) (Over the Packed Bed During Operation)
22 psi
bed size
16 mm × 100 mm
bed volume
20 mL
column I.D.
16 mm
matrix
6% cross-linked agarose
particle size
45-165 μm
average diameter
90 μm
cleaning
2-14(Ni2+-stripped medium.)
working range
3-12(Ni2+-stripped medium.)
capacity
~40 mg binding capacity(histidine-tagged protein)
suitability
suitable for bioprocess medium
Related Categories
General description
HisPrep™ FF 16/10 columns are prepacked with Ni Sepharose™ 6 Fast Flow for scale-up purification of histidine-tagged proteins. Ni Sepharose™ 6 Fast Flow consists of 90 ?m beads of highly cross-linked agarose, to which a chelating ligand has been immobilized. This chelating ligand is charged with Ni2+ ions, the first-choice metal ion for purifying most histidine-tagged proteins. The negligible leakage of Ni2+ ions ensures reliable capture of histidine-tagged proteins in repeated IMAC purifications.
Features and Benefits
- Optimized for convenient scale-up purification of histidine-tagged proteins.
- Compatible with a wide range of reducing agents, detergents, denaturants, and other additives.
- Negligible leakage of Ni2+.
- High binding capacity, approx. 40 mg/mL medium.
- Convenient and time-saving 20 mL prepacked HiPrep™ format.
Storage and Stability
Analysis Note
Other Notes
Legal Information
Signal Word
Warning
Hazard Statements
Precautionary Statements
Storage Class Code
3 - Flammable liquids
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Articles
This page shows how to purify or remove proteins and peptides with exposed amino acids with Chelating Sepharose High Performance, Chelating Sepharose Fast Flow, Capto Chelating from Cytiva.
This page shows how to perform a purification and on-column refolding of an insoluble his-tagged protein from an E. coli culture with HisTrap™ FF columns and ÄKTAprime from Cytiva.
This page shows the characteristics of Ni Sepharose, Ni Sepharose excel, TALON Superflow, and uncharged IMAC Sepharose products from Cytiva.
This page shows how to convert between flow velocity and volumetric flow rate in affinity chromatography of antibodies.
Protocols
Ni Sepharose 6 Fast Flow purifies histidine-tagged proteins efficiently, offering high cross-linked agarose beads with Ni2+ ions.
Sample Preparation for Affinity Chromatography in Specific Groups of Biomolecules
This page shows how to perform sample desalting, buffer exchange and concentration for affinity chromatography of tagged proteins.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service