Skip to Content
Merck
All Photos(1)

Key Documents

GE17-5268-01

Ni Sepharose High Performance

Cytiva 17-5268-01, pack of 25 mL

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106500
NACRES:
NA.56
Pricing and availability is not currently available.

packaging

pack of 25 mL

manufacturer/tradename

Cytiva 17-5268-01

matrix

highly cross-linked 6% agarose

avg. part. size

34 μm

cleaning in place

2-14(Ni2+-stripped medium.)

working range

3-12(Ni²⁺-stripped medium.)

capacity

≥40 mg binding capacity (histidine-tagged protein/ml medium)(Protein binding capacity is protein-to-protein dependent.)

General description

Ni Sepharose High Performance affinity media for high-resolution, high capacity purification of histidine-tagged proteins by immobilized metal ion affinity chromatography (IMAC).

Ni Sepharose High Performance consists of highly cross-linked agarose beads to which a chelating group has been coupled. This chelating group is precharged with Ni2+, which will selectively retain proteins with exposed histidine groups.

The high stability and broad compatibility of Ni Sepharose High Performance maintains biological activity and increases product yield, at the same time as it greatly expands the range of suitable operating conditions.

Features and Benefits

  • High-performance purification of histidine-tagged proteins
  • Negligible leakage of Ni2+ ions.
  • High binding capacity, at least 40 mg/mL medium
  • Compatible with a wide range of reducing agents, detergents, denaturants and other additives
  • 34 μm bead size for improved resolution

Storage and Stability

Please be aware this product may be shipped 90 days before the expiration date. For more information on the batch specific expiration date, please contact technical service.
Store at 4 to 30 °C (20% Ethanol)

Analysis Note

To view the Certificate of Analysis for this product, please visit www.cytiva.com.

Legal Information

IMAC Sepharose products, Ni Sepharose products and Fe Sepharose products
These products are sold under a license from Sigma-Aldrich under patent number EP 1276716 (Metal chelating compositions) and equivalent patents and patent applications in other countries.
Sepharose is a trademark of Cytiva

Pictograms

Environment

Signal Word

Warning

Hazard Statements

Storage Class Code

3 - Flammable liquids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

This page shows how to purify or remove proteins and peptides with exposed amino acids with Chelating Sepharose High Performance, Chelating Sepharose Fast Flow, Capto Chelating from Cytiva.

This page shows high-throughput screening using His MultiTrap™ HP and His MultiTrap™ FF 96-well filter plates from Cytiva.

Follow established procedures for recombinant protein expression, growth, induction, and cell lysis optimization.

Protocols

This page describes a small-scale purification (minipreps) of histidine-tagged recombinant proteins using His SpinTrap and His SpinTrap Kit from Cytiva.

Ni Sepharose High Performance consists of highly cross-linked 6% agarose beads (34 µm) to which a chelating group has been immobilized and subsequently charged with Ni2+ ions.

This page shows how to purify protein A-tagged proteins using Cytiva products.

How to isolate proteins from inclusion bodies using Cytiva products.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service