Skip to Content
Merck
All Photos(1)

Documents

GE28-4065-57

dNTP Set - A, C, G, T

Cytiva 28-4065-57, pack of 1 × 10 μmol

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106305
NACRES:
NA.52

form

liquid

packaging

pack of 1 × 10 μmol

manufacturer/tradename

Cytiva 28-4065-57

concentration

20 mM

color

Colorless

shipped in

dry ice

storage temp.

−20°C

Related Categories

General description

DNA Polymerization Mix dNTP Set (20 mM each A,C,G,T).

Application

By incorporating recent advances in manufacturing technology, Cytiva now offers illustra Solution dNTPs: the highest purity deoxynucleotides available for amplification, dideoxy sequencing, labeling, mutagenesis, cDNA synthesis, and expression profiling.

Features and Benefits

  • Free from DNase, RNase, and nicking enzyme activity
  • Greater than 99% triphosphate purity (by HPLC) for optimum performance and consistency
  • Buffer free and ready-to-use solutions in multiple formats
  • Functionally tested for long PCR* and sequencing
  • Custom formulation, testing, packaging, and labeling are available

Storage and Stability

Please be aware this product may be shipped 90 days before the expiration date. For more information on the batch specific expiration date, please contact technical service.

Analysis Note

To view the Certificate of Analysis for this product, please visit www.cytiva.com.

Legal Information

illustra is a trademark of Cytiva

Storage Class Code

12 - Non Combustible Liquids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service