Skip to Content
Merck
All Photos(1)

Key Documents

8890-OP

Millipore

TE Buffer Solution pH 8.0

Synonym(s):

1X TE buffer, Tris EDTA buffer

Sign Into View Organizational & Contract Pricing

Select a Size

100 ML
Inquire


Estimated to ship on25 March 2025


Request a Bulk Order

Select a Size

Change View
100 ML
Inquire

About This Item

UNSPSC Code:
41105319
NACRES:
NA.21


Estimated to ship on25 March 2025


Request a Bulk Order

form

solution

Quality Level

manufacturer/tradename

Calbiochem®

shipped in

ambient

General description

1X TE Solution, pH 8.0 (10mM TRIS-HCl, 1mM EDTA). Molecular Biology grade.

Specifications:
Conductivity at 25°C: 775-975 µmho
DNase: None detected
pH of solution at 25°C: 7.90-8.10
Protease: None detected
RNase: None detected





Intended for laboratory and manufacturing use only. Not for drug, food, or household use.
Tris- ethylenediaminetetraacetic (TE) buffer is generally used in molecular biology as a storage solution to maintain the integrity of nucleic acids.[1]

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Corrosion

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Met. Corr. 1 - Skin Irrit. 2

Storage Class Code

8B - Non-combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jason W Johnston et al.
Plant physiology and biochemistry : PPB, 43(9), 844-853 (2005-11-18)
HPLC analysis of nucleosides is important for determining total DNA methylation in plants and can be used to help characterise epigenetic changes during stress, growth and development. This is of particular interest for in vitro plant cultures as they are

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service