Skip to Content
Merck
All Photos(2)

Key Documents

GBL615101

Grace Bio-Labs HybriWell seals, assortment

with applicator stick, pack of 8 ea

Synonym(s):

chamber slides, chambered coverglass, cover slides, hybridization chambers, incubation chambers

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
€226.00

€226.00


Check Cart for Availability


Select a Size

Change View
1 EA
€226.00

About This Item

UNSPSC Code:
41122600
NACRES:
NB.22

€226.00


Check Cart for Availability

material

silicone (on glass)

sterility

non-sterile

feature

wells 1

packaging

pack of 8 ea

manufacturer/tradename

Grace Bio-Labs 615101

external size

Various mm × Various mm

General description

A Selection of Reaction Chambers that Every Lab Should Have on Hand

Conserve precious samples and reagents by selecting the optimal area and volume for your application and create a secure seal to eliminate evaporation. HybriWells are easy-to-use enclosures for use on a variety of smooth surfaces. Disposable chamber removes cleanly and easily, even after heating. HybriWell seals are heat stable to 97 °C, and are resistant to aqueous buffers and detergents. Made of nuclease-free, hydrophobic material the HybriWell will not trap or bind probes and is completely compatible with biological samples.

Application includes:
  • In situ hybridization
  • Microarrays
  • Immunocytochemistry
  • DNA Amplification
  • Neurophysiology
  • Reaction kinetics
  • Rapid microfluidic protoyping
  • Nucleic Acid Programmable Protein Arrays

Legal Information

HybriWell is a trademark of Grace Bio-Labs, Inc.

Kit Components Also Available Separately

Product No.
Description
SDS

  • GBL611101Grace Bio-Labs HybriWell sealing system, SecureSeal adhesive chamber, chamber volume 18 μL, diam. × thickness 13 mm × 0.15 mm, port diam. 1.5 mmSDS

  • GBL611102Grace Bio-Labs HybriWell sealing system, SecureSeal adhesive chamber, chamber volume 30 μL, diam. × thickness 20 mm × 0.15 mm, port diam. 1.5 mmSDS

  • GBL611104Grace Bio-Labs HybriWell sealing system, SecureSeal adhesive chamber, chamber volume 30-50 μL, L × W × thickness 21.6 mm × 21.6 mm × 0.15 mm, port diam. 1.5 mmSDS

  • GBL611105Grace Bio-Labs HybriWell sealing system, SecureSeal adhesive chamber, chamber volume 50-100 μL, L × W × thickness 21 mm × 40 mm × 0.15 mm, port diam. 3.2 mmSDS

  • GBL611205Grace Bio-Labs HybriWell-FL sealing system, Fluor-"friendly" adhesive chamber, chamber volume 50-100 μL, L × W × thickness 22 mm × 40 mm × 0.12 mm, port diam. 1.5 mmSDS

  • GBL612105Grace Bio-Labs HybriWell sealing system, SecureSeal adhesive chamber, chamber volume 275-320 μL, L × W × thickness 60 mm × 21 mm × 0.15 mm, port diam. 3.2 mmSDS

  • GBL612101Grace Bio-Labs HybriWell sealing system, SecureSeal adhesive chamber, chamber volume 180-200 μL, L × W × thickness 22 mm × 40 mm × 0.25 mm, port diam. 1.5 mmSDS

  • GBL612202Grace Bio-Labs HybriWell-FL sealing system, Fluor-"friendly" adhesive chamber, chamber volume 280-330 μL, L × W × thickness 22 mm × 60 mm × 0.25 mm, port diam. 1.5 mmSDS

See All (8)

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service