Skip to Content
Merck
All Photos(2)

Key Documents

GBL611102

Grace Bio-Labs HybriWell sealing system, SecureSeal adhesive chamber

chamber volume 30 μL, diam. × thickness 20 mm × 0.15 mm, port diam. 1.5 mm

Synonym(s):

chamber slides, chambered coverglass, cover slides, hybridization chambers, incubation chambers

Sign Into View Organizational & Contract Pricing

Select a Size

100 EA
€301.00

€301.00


Check Cart for Availability


Select a Size

Change View
100 EA
€301.00

About This Item

UNSPSC Code:
41122600
NACRES:
NB.22

€301.00


Check Cart for Availability

material

colorless
polycarbonate

sterility

non-sterile

feature

wells 1

packaging

pack of 100 ea

manufacturer/tradename

Grace Bio-Labs 611102

chamber volume

30 μL

diam. × thickness

20 mm × 0.15 mm

port diam.

1.5 mm

wells

1

Looking for similar products? Visit Product Comparison Guide

General description

Grace Bio-Labs HybriWell sealing system seals securely to a microscope slide surface in seconds to accommodate waterbath hybridizations, such as protein or nucleic acid hybridization assays.

Features:
  • Encloses a single or multiple specimens in a small reagent volume and eliminate evaporation
  • Access ports in the chamber surface allow for the addition or removal of reactants
  • Ports are easily sealed using adhesive seal tabs provided with each order
  • Sealed chambers are watertight and ideally suited for waterbath incubations
  • RNase free, hydrophobic surfaces will not trap or bind probes
  • Disposable chamber removes cleanly and easily even after heating
  • Each package includes illustrated instructions for use, applicator and 200 adhesive port seals

Legal Information

HybriWell is a trademark of Grace Bio-Labs, Inc.
SecureSeal is a trademark of Grace Bio-Labs, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service