Skip to Content
Merck
All Photos(2)

Key Documents

GBL246871

Grace Bio-Labs silicone gasket for ProPlate® microarray system

fits 1 well, size 16.5 mm × 70 mm

Synonym(s):

microarray chambers for slides, silicone slide gasket

Sign Into View Organizational & Contract Pricing

Select a Size

10 EA
€71.60

€71.60


Check Cart for Availability


Select a Size

Change View
10 EA
€71.60

About This Item

UNSPSC Code:
41122600
NACRES:
NB.15

€71.60


Check Cart for Availability

sterility

non-sterile

feature

wells 1

packaging

pack of 10 ea

manufacturer/tradename

Grace Bio-Labs 246871

size

1 well
16.5 mm × 70 mm

color

none

Looking for similar products? Visit Product Comparison Guide

General description

Replacement Silicone Gaskets for Grace BioLabs Proplate slide modules.

While the silicone gaskets affixed to wells may be re-used multiple times, their lifespan may be shortened by washing conditions, repeated autoclaving and prolonged exposure to harsher types of chemicals. Therefore, replacement gaskets are available and may be easily positioned and applied to the chambers after simply peeling the old gasket off and throwing it away.

Application

  • Protein and peptides microarrays
  • cDNA and oligonucleotides microarrays
  • Proteomics
  • Protein expression analysis
  • Antibody profiling

Legal Information

ProPlate is a registered trademark of Grace Bio-Labs, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service