Skip to Content
Merck
All Photos(2)

Key Documents

GBL204860

Grace Bio-Labs ProPlate® tray sets

size 16 wells, with Delrin® snap clips, well L × W 7 mm × 7 mm

Synonym(s):

incubation chambers, microarray hybridization chambers, slide chambers

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
€271.00

€271.00


Check Cart for Availability


Select a Size

Change View
1 EA
€271.00

About This Item

UNSPSC Code:
41122800
NACRES:
NB.15
size:
16 wells

€271.00


Check Cart for Availability

sterility

non-sterile

feature

wells 16

packaging

set of 1

manufacturer/tradename

Grace Bio-Labs 204860

size

16 wells

well L × W

7 mm × 7 mm

well volume

350 μL

color

none

Looking for similar products? Visit Product Comparison Guide

General description

Grace Bio-Labs ProPlate® multi-array chamber tray sets are designed to be used with ONCYTE® nitrocellulose-coated slides. These multi-array chambers integrate microscope slide based microarray technology with high-throughput (HTS applications) microtiter plate processing. Up to 64 individual arrays can be assayed per slide.

Three or four slides fit into a tray (depending on clip choice). The resulting multi-array chamber tray set produces a modular plate with a standard microtiter plate footprint and well spacing allowing high-throughput robotic processing of up to 256 arrays.

Features:

  • Up to 16 or 64 leak-proof wells on any slide surface without the use of adhesive
  • Microtiter plate well spacing facilitates use with multichannel pipettes
  • 7 x 7 mm square wells are ideal for high content arrays 5-350 uL well volumes accommodate small reagent and large wash volumes

16 or 64 well ProPlate® tray sets include the following:

  • 7 x 7 mm well dimensions (x 4)
  • 16 or 64 well capacity
  • Stainless steel Delrin® snap clips (x 8)
  • Tray & cover included (x 1)
  • 10 seal strips (x 10) and 1 seal strip applicator (x 1)

Legal Information

Delrin is a registered trademark of E. I. du Pont de Nemours and Company
Oncyte is a registered trademark of Grace Bio-Labs, Inc.
ProPlate is a registered trademark of Grace Bio-Labs, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service