Skip to Content
Merck
All Photos(2)

Key Documents

GBL204874

Grace Bio-Labs SealStrips and SealStrip applicator, for ProPlate® microarray systems

for ProPlate® microarray systems, pkg of 50 ea

Synonym(s):

GRACE BIO-LABS SEALSTRIPS AND SEALSTRIP APPLICATOR, FOR PROPLATE® MICROARRAY SYSTEMS

Sign Into View Organizational & Contract Pricing

Select a Size

50 EA
€66.70

€66.70


Estimated to ship on15 April 2025



Select a Size

Change View
50 EA
€66.70

About This Item

UNSPSC Code:
41121800
NACRES:
NB.15

€66.70


Estimated to ship on15 April 2025


sterility

non-sterile

packaging

pkg of 50 ea

manufacturer/tradename

Grace Bio-Labs 204874

external size

18 mm × 77 mm

color

none

Looking for similar products? Visit Product Comparison Guide

General description

The adhesive strips being offered are made from materials that are commonly used for sealing PCR plates. They are compatible with hybridization reagents and can be used without causing any contamination. The pack includes an applicator and 50 seal strips. The seal strips should be applied on the top surface of the chamber using the plastic applicator to avoid evaporation. When removing the strips, they come off easily and cleanly, leaving no unwanted residue.

Legal Information

ProPlate is a registered trademark of Grace Bio-Labs, Inc.
SealStrips is a trademark of Grace Bio-Labs, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service