Skip to Content
Merck
All Photos(1)

Documents

G8171

Sigma-Aldrich

Gambogic acid

≥95% (HPLC), powder

Synonym(s):

1,5-Methano-1H,3H,11H-furo(3,4-g)pyrano(3,2-b)xanthene-1-crotonic acid, B"-Guttiferrin, Beta-Guttiferrin, Beta-Guttilactone, Cambogic acid, Guttatic acid, Guttic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C38H44O8
CAS Number:
Molecular Weight:
628.75
UNSPSC Code:
12352200
PubChem Substance ID:

Assay

≥95% (HPLC)

form

powder

color

yellow to orange

solubility

DMSO: ≥10 mg/mL
H2O: insoluble

storage temp.

−20°C

SMILES string

C\C(C)=C\CC[C@@]1(C)Oc2c(C\C=C(\C)C)c3O[C@@]45[C@@H]6C[C@@H](C=C4C(=O)c3c(O)c2C=C1)C(=O)[C@]5(C\C=C(\C)C(O)=O)OC6(C)C

InChI

1S/C38H44O8/c1-20(2)10-9-15-36(8)16-14-24-29(39)28-30(40)26-18-23-19-27-35(6,7)46-37(33(23)41,17-13-22(5)34(42)43)38(26,27)45-32(28)25(31(24)44-36)12-11-21(3)4/h10-11,13-14,16,18,23,27,39H,9,12,15,17,19H2,1-8H3,(H,42,43)/b22-13-/t23-,27+,36-,37+,38-/m1/s1

InChI key

GEZHEQNLKAOMCA-RRZNCOCZSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Gambogic acid acts as a caspase activator and apoptosis inducer, which causes an irreversible arrest in the G2/M phase of the cell cycle.

Features and Benefits

This compound is featured on the Caspases page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jing Geng et al.
Inflammation research : official journal of the European Histamine Research Society ... [et al.], 62(2), 165-172 (2012-10-16)
Gambogic acid (GBA) targeted Heat shock protein 90 (Hsp90) and prohibited TNF-α/NF-κB signaling pathway. It can be inferred that the anti-inflammatory activity of GBA results from inhibiting the cytokine production via NF-κB signaling pathway. We used the RAW264.7 cell line
Jing Yang et al.
Journal of natural products, 75(6), 1108-1116 (2012-06-06)
Gambogic acid (1) is a cytotoxic caged xanthone derived from the resin of Garcinia hanburyi. Compound 1 selectively induces apoptosis in cancer cells, at least partially, by targeting the stress response to reactive oxygen species (ROS). However, the molecular mechanism
Hao-Peng Sun et al.
Chemistry & biodiversity, 9(8), 1579-1590 (2012-08-18)
The natural product gambogic acid exhibits high potency in inhibiting cancer cell lines. Rational medicinal modifications on gambogic acid will improve its physicochemical properties and drug-like characters. To investigate the structure-activity relationship of gambogic acid and also to find rational
Xu Wang et al.
Anti-cancer agents in medicinal chemistry, 12(8), 994-1000 (2012-02-22)
Gambogic acid (GA) is a caged xanthone that is derived from Garcinia hanburyi and functions as a strong apoptotic inducer in many types of cancer cells. The distinct effectiveness of GA has led to its characterization as a novel anti-cancer
Gianni Guizzunti et al.
Chembiochem : a European journal of chemical biology, 13(8), 1191-1198 (2012-04-26)
The natural product gambogic acid (GA) has shown significant potential as an anticancer agent as it is able to induce apoptosis in multiple tumor cell lines, including multidrug-resistant cell lines, as well as displaying antitumor activity in animal models. Despite

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service