Skip to Content
Merck
All Photos(1)

Key Documents

345701

Sigma-Aldrich

Gambogic Acid, Garcinia hanburyi

A cell-permeable caspase activator and an apoptosis inducer that was originally isolated from gamboge resin for its antimicrobial properties.

Synonym(s):

Gambogic Acid, Garcinia hanburyi, Guttic Acid, β-Guttiferin

Sign Into View Organizational & Contract Pricing

Select a Size

25 MG
€298.00

€298.00


Estimated to ship on24 May 2025


Request a Bulk Order

Select a Size

Change View
25 MG
€298.00

About This Item

Empirical Formula (Hill Notation):
C38H44O8
CAS Number:
Molecular Weight:
628.75
MDL number:
UNSPSC Code:
12352106
NACRES:
NA.77

€298.00


Estimated to ship on24 May 2025


Request a Bulk Order

Quality Level

description

Merck USA index - 14, 4361

Assay

≥95% (TLC)

form

amorphous solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

orange

solubility

DMSO: 10 mg/mL
ethanol: soluble

shipped in

ambient

storage temp.

2-8°C

SMILES string

O1[C@@]32C4(OC(C3CC(C=C2C(=O)c5c1c(c6c(c5O)C=C[C@](O6)(CCC=C(C)C)C)CC=C(C)C)C4=O)(C)C)C\C=C(\C)/C(=O)O

InChI

1S/C38H44O8/c1-20(2)10-9-15-36(8)16-14-24-29(39)28-30(40)26-18-23-19-27-35(6,7)46-37(33(23)41,17-13-22(5)34(42)43)38(26,27)45-32(28)25(31(24)44-36)12-11-21(3)4/h10-11,13-14,16,18,23,27,39H,9,12,15,17,19H2,1-8H3,(H,42,43)/b22-13-/t23?,27?,36-,37?,38-/m1/s1

InChI key

GEZHEQNLKAOMCA-PLUQQRNKSA-N

General description

A cell-permeable caspase activator and an apoptosis inducer that was originally isolated from gamboge resin for its antimicrobial properties. Shown to inhibit the growth of HeLa and HEL cells minimal inhibitory concentrations (MIC = 12.5 µg/ml).
A cell-permeable, anti-microbial xanthone orignally isolated from the gamboge resin of Garcinia hanburyi. Acts as a caspase activator and apoptosis inducer. Shown to inhibit the growth of HeLa and HEL cells (MIC = 12.5 µg/ml). Reported to block telomerase activity in SMMC-7721 cells. Shown to induce tyrosine kinase phosphatase SHP-1 and suppress Stat3 activation.

Biochem/physiol Actions

Cell permeable: yes
MIC = 12.5 µg/ml in inhibiting the growth of HeLa and HEL cells
Primary Target
Caspase activator and an apoptosis inducer
Product does not compete with ATP.
Reversible: no

Packaging

Packaged under inert gas

Warning

Toxicity: Toxic & Carcinogenic / Teratogenic (G)

Other Notes

Due to the nature of the Hazardous Materials in this shipment, additional shipping charges may be applied to your order. Certain sizes may be exempt from the additional hazardous materials shipping charges. Please contact your local sales office for more information regarding these charges.
Prasad, S., et al. 2011. Cancer Prev. Res.4, 1084.
Guo, Q.L., et al. 2004. Acta Pharmacol. Sin.25, 769.
Asano, J., et al. 1996. Phytochemistry41, 815.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Qingwen Zhu et al.
Autophagy, 17(7), 1667-1683 (2020-07-07)
Dysregulated microRNAs (miRNAs) are involved in carcinoma progression, metastasis, and poor prognosis. We demonstrated that in nasopharyngeal carcinoma (NPC), transactivated MIR106A-5p promotes a malignant phenotype by functioning as a macroautophagy/autophagy suppressor by targeting BTG3 (BTG anti-proliferation factor 3) and activating

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service